Skip to Content
Merck
All Photos(1)

Key Documents

440272

Sigma-Aldrich

Dichlorodimethylsilane

Dichlorodimethylsilane

≥99.5%

Synonym(s):

DMDCS, Dimethyldichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SiCl2
CAS Number:
Molecular Weight:
129.06
Beilstein:
605287
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥99.5%

form

liquid

refractive index

n20/D 1.404 (lit.)

bp

70 °C (lit.)

mp

−76 °C (lit.)

density

1.07 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(Cl)Cl

InChI

1S/C2H6Cl2Si/c1-5(2,3)4/h1-2H3

InChI key

LIKFHECYJZWXFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Common reagent for synthesis of optically active ansa-mettallocene polymerization catalysts.[1]
Used to prepare a resin bound siloxane with reactivity towards tertiary alcohols.[2]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

33.8 °F - closed cup

Flash Point(C)

1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 63, 299-299 (2007)
Cliff R Baar et al.
Journal of the American Chemical Society, 126(26), 8216-8231 (2004-07-01)
A series of enantiopure C1-symmetric metallocenes, [(SiMe2)2[eta5-C5H(CHMe2)2][eta5-C5H2((S)-CHMeCMe3)]]ZrCl2, (S)-2, [(SiMe2)2[eta5-C5H(CHEt2)2][eta5-C5H2((S)-CHMeCMe3)]]ZrCl2, (S)-6, and [(SiMe2)2[eta5-C5HCy2][eta5-C5H2((S)-CHMeCMe3)]]ZrCl2, (S)-7 (Cy = cyclohexyl), zirconocene dichlorides that have an enantiopure methylneopentyl substituent on the "upper" cyclopentadienyl ligand, and diastereomerically pure precatalysts, [(SiMe2)2[eta5-C5H((S)-CHMeCy)(CHMe2)][eta5-C5H3]]ZrCl2, (S)-8a and (S)-8b, which have an
J M Curran et al.
Biomaterials, 32(21), 4753-4760 (2011-04-15)
Material modifications can be used to induce cell responses, in particular-CH(3) and -NH(2) have shown potential in enhancing the ability of a material to support mesenchymal stem cell (MSC) adhesion and differentiation. Currently this process is variable, due to the
K E Healy et al.
Biomaterials, 17(2), 195-208 (1996-01-01)
Materials with spatially resolved chemistries (i.e. patterned surfaces) have been used to guide and organize the position of mammalian cells in vitro. A common theme in guiding the spatial distribution of cells has been the use of patterned alkylsiloxanes, where
Y C Tseng et al.
Biomaterials, 16(13), 963-972 (1995-09-01)
Amphipathic ethylene glycol-butadiene block copolymers (PEG-PB) with different chain lengths of poly(ethylene glycol) (PEG) were synthesized by reacting poly(ethylene glycol methyl ether) (m-PEG, mol. wt = 350, 550, 750, 2000 and 5000) with telechelic polybutadiene (PB). The PEG-PB copolymers formed

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Is item 440272, Dichlorodimethylsilane, in a sure seal bottle?

    1 answer
    1. The PHBOM5691 - 440272, Dichlorodimethylsilane, comes in a 100ml size with a sure seal.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service