Skip to Content
Merck
All Photos(2)

Key Documents

144207

Sigma-Aldrich

Chlorodimethylsilane

98%

Synonym(s):

DMCS, Dimethylchlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SiHCl
CAS Number:
Molecular Weight:
94.62
Beilstein:
1633434
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

vapor density

4 (vs air)

vapor pressure

8.56 psi ( 20 °C)

Assay

98%

form

liquid

expl. lim.

20 %

refractive index

n20/D 1.383 (lit.)

bp

34.7 °C (lit.)

mp

−111 °C (lit.)

density

0.852 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[SiH](C)Cl

InChI

1S/C2H7ClSi/c1-4(2)3/h4H,1-2H3

InChI key

YGHUUVGIRWMJGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used to prepare vinyldimethylsilanols for palladium-catalyzed cross-coupling with aryl iodides.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Flam. Liq. 1 - Skin Corr. 1A - Water-react 3

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

-9.4 °F - closed cup

Flash Point(C)

-23 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yue Huang et al.
Langmuir : the ACS journal of surfaces and colloids, 35(30), 9785-9793 (2019-07-10)
A series of sulfonate-based silicone surfactants with different hydrophobic groups were synthesized. Two synthetic strategies are introduced to permit exquisite control over the hydrophobic moieties. Solution behavior of these surfactants was investigated by surface tensiometry, electrical conductivity, transmission electron microscopy
Total synthesis of papulacandin D.
Scott E Denmark et al.
Journal of the American Chemical Society, 129(10), 2774-2776 (2007-02-20)
Kirti Ramesh et al.
Nature communications, 8(1), 1709-1709 (2017-11-24)
Understanding mollusk calcification sensitivity to ocean acidification (OA) requires a better knowledge of calcification mechanisms. Especially in rapidly calcifying larval stages, mechanisms of shell formation are largely unexplored-yet these are the most vulnerable life stages. Here we find rapid generation
Paulina Teper et al.
Polymers, 12(1) (2020-01-23)
In this paper, we focus on the synthesis and characterization of novel stable nanolayers made of star methacrylate polymers. The effect of nanolayer modification on its antibacterial properties was also studied. A covalent immobilization of star poly(N,N'-dimethylaminoethyl methacrylate) (PDMAEMA) to
S Evans et al.
Environmental pollution (Barking, Essex : 1987), 230, 368-377 (2017-07-04)
Due to concerns regarding the release of pharmaceuticals into the environment and the understudied impact of stereochemistry of pharmaceuticals on their fate and biological potency, we focussed in this paper on stereoselective transformation pathways of selected chiral pharmaceuticals (16 pairs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service