Skip to Content
Merck
All Photos(1)

Key Documents

439665

Sigma-Aldrich

Strontium chloride

anhydrous, powder, ≥99.99% trace metals basis

Synonym(s):

Strontium dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrCl2
CAS Number:
Molecular Weight:
158.53
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
anhydrous
form:
powder

grade

anhydrous

Assay

≥99.99% trace metals basis

form

powder

impurities

≤100.0 ppm Trace Metal Analysis

mp

874 °C (lit.)

density

3 g/mL at 25 °C (lit.)

SMILES string

Cl[Sr]Cl

InChI

1S/2ClH.Sr/h2*1H;/q;;+2/p-2

InChI key

AHBGXTDRMVNFER-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Strontium chloride hexahydrate (SrCl2·6H2O) is a hydrated alkaline earth metal chloride. On γ-irradiation at 77°K, it affords a radical having e.s.r. parameters resembling with HO2. Its chlorine electric field gradient (EFG) and chemical shift (CS) tensors have been evaluated by employing solid-state 35/37Cl NMR spectroscopy.

Application

The product was used as a source of Sr2+ ions, to evaluate the working of a Flow Electrolysis Cell used in Rapid Ion Transfer (FECRIT) electrochemical solvent extraction method. Supernatant solutions of SrCl2 may also be used as supplier of Sr to prepare fibrous strontium substituted hydroxyapatite, a potential reinforcement for biocomposites.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Micha Ben-Naim et al.
ACS applied materials & interfaces, 12(5), 5901-5908 (2020-01-24)
Silicon has shown promise for use as a small band gap (1.1 eV) absorber material in photoelectrochemical (PEC) water splitting. However, the limited stability of silicon in acidic electrolyte requires the use of protection strategies coupled with catalysts. Herein, spin
Fibrous growth of strontium substituted hydroxyapatite and its drug release.
Suganthi RV, et al.
Materials Science & Engineering. C, Materials For Biological Applications, 31(3), 593-599 (2011)
Development of high performance electrochemical solvent extraction method
Okugaki T, et al.
Journal of Electroanalytical Chemistry, 629(1-2), 50-56 (2009)
Unstable intermediates. Part LX. HO2 radical in γ-irradiated strontium chloride hexahydrate.
Catton RC and Symons MCR.
J. Chem. Soc. Sect. A, 1393-1395 (1969)
T C Brennan et al.
British journal of pharmacology, 157(7), 1291-1300 (2009-07-01)
Strontium ranelate reduces fracture risk in postmenopausal women with osteoporosis. Evidence from non-clinical studies and analyses of bone markers in phase III trials indicate that this is due to an increase in osteoblast formation and a decrease of osteoclastic resorption.

Articles

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service