204463
Strontium chloride hexahydrate
99.995% trace metals basis
Synonym(s):
Strontium dichloride hexahydrate
About This Item
Assay
99.995% trace metals basis
form
powder and chunks
impurities
≤55.0 ppm Trace Metal Analysis
mp
115 °C (lit.)
solubility
ethanol: slightly soluble(lit.)
SMILES string
O.O.O.O.O.O.Cl[Sr]Cl
InChI
1S/2ClH.6H2O.Sr/h2*1H;6*1H2;/q;;;;;;;;+2/p-2
InChI key
AMGRXJSJSONEEG-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Application
- Development of novel composite PCM for thermal energy storage using CaCl2· 6H2O with graphene oxide and SrCl2· 6H2O: This study details the development of a new composite phase change material incorporating strontium chloride hexahydrate, relevant for material scientists focused on energy storage solutions (Xu et al., 2017).
- Toxicity assessment of artificially added zinc, selenium, and strontium in water: This research assesses the toxicity of strontium chloride hexahydrate in aquatic environments, which is crucial for environmental chemists and toxicologists studying the impact of chemical agents on aquatic life (Liu et al., 2019).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service