Skip to Content
Merck
All Photos(1)

Key Documents

439657

Sigma-Aldrich

Terbium(III) chloride

anhydrous, powder, 99.9% trace metals basis

Synonym(s):

Terbium chloride, Terbium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TbCl3
CAS Number:
Molecular Weight:
265.28
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
anhydrous
form:
powder

grade

anhydrous

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: terbium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

mp

588 °C (lit.)

density

4.35 g/mL at 25 °C (lit.)

SMILES string

Cl[Tb](Cl)Cl

InChI

1S/3ClH.Tb/h3*1H;/q;;;+3/p-3

InChI key

GFISHBQNVWAVFU-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Shinohara et al.
Toxicology, 66(1), 93-103 (1991-02-11)
To investigate the biological effects of terbium (Tb), male mice were intraperitoneally administered with TbCl3 at doses of 10, 50, or 250 mg Tb/kg. The Tb distribution in organs was determined after 18-20 h of injection by using a spectrofluorometer.
H Shimada et al.
Journal of toxicology and environmental health, 48(1), 81-92 (1996-05-01)
Terbium (Tb) is a rare earth metal that finds use in several emerging technologies. However, little is known about the biological effects of Tb. Thus, in this study the pulmonary toxicity of systemic Tb in mice was investigated. Mice were
J L Cox et al.
Melanoma research, 12(3), 211-219 (2002-07-26)
Alterations in the extracellular Ca(2+) or K(+) concentration had significant influences on the motility of B16F10 melanoma cells measured in the absence of exogenous integrins using a conventional Boyden chamber assay. At normal K(+) concentrations, motility increased slightly when the
William J Rieter et al.
Journal of the American Chemical Society, 130(35), 11584-11585 (2008-08-09)
Pt-containing nanoscale coordination polymer (NCP) particles with the formula of Tb2(DSCP)3(H2O)12 (where DSCP represents disuccinatocisplatin), NCP-1, were precipitated from an aqueous solution of Tb3+ ions and DSCP bridging ligands via the addition of a poor solvent. SEM and TEM images
T Pineda et al.
Biochimica et biophysica acta, 1292(2), 259-264 (1996-02-08)
myo-Inositol monophosphatase catalyzes dephosphorylation of the synthetic substrate anthraniloyl-2'-AMP. Binding of this fluorescent substrate to Tb(III)-monophosphatase was monitored by luminescence spectroscopy. The anthraniloyl chromophore excited at 330 nm sensitizes the long lived luminescence of enzyme bound Tb(III) at 490, 545

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service