Skip to Content
Merck
All Photos(1)

Key Documents

43116

Sigma-Aldrich

N-(Diphenylmethyl)methylamine

≥98.0% (GC)

Synonym(s):

N-Methylbenzhydrylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15N
CAS Number:
Molecular Weight:
197.28
Beilstein:
2096409
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

form

solid

mp

38-42 °C

functional group

amine
phenyl

SMILES string

CNC(c1ccccc1)c2ccccc2

InChI

1S/C14H15N/c1-15-14(12-8-4-2-5-9-12)13-10-6-3-7-11-13/h2-11,14-15H,1H3

InChI key

SHDMMLFAFLZUEV-UHFFFAOYSA-N

General description

N-(Diphenylmethyl)methylamine is a sterically hindered secondary amine. It participates in the synthesis of cyclooctene oxide.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-Diphenylmethyl-2-propenamide: theoretical study of the structure and interaction with a DNA model system.
Barone G, et al.
Journal of Molecular Structure, 572(1), 113-119 (2001)
Ganesan Rajasekaran et al.
Biochimica et biophysica acta, 1859(5), 722-733 (2017-02-06)
Although the human-derived antimicrobial peptide (AMP) LL-37 has potent antimicrobial and anti-inflammatory activities, its therapeutic application is limited by its low cell selectivity and high production cost due to its large size. To overcome these problems, we tried to develop
Svetlana N Rylova et al.
PloS one, 13(4), e0195802-e0195802 (2018-04-19)
Copper-64 is an attractive radionuclide for PET imaging and is frequently used in clinical applications. The aim of this study was to perform a side-by-side comparison of the in vitro and in vivo performance of 64Cu-NODAGA-JR11 (NODAGA = 1,4,7-triazacyclononane,1-glutaric acid,4,7-acetic
Eun Young Kim et al.
European journal of medicinal chemistry, 136, 428-441 (2017-05-20)
KR-12-a5 is a 12-meric α-helical antimicrobial peptide (AMP) with dual antimicrobial and anti-inflammatory activities designed from human cathelicidin LL-37. We designed and synthesized a series of d-amino acid-substituted analogs of KR-12-a5 with the aim of developing novel α-helical AMPs that
He Ma et al.
ChemMedChem, 14(19), 1710-1716 (2019-08-25)
Naturally occurring constrained peptides are frequently used as scaffolds for bioactive peptide grating due to their high stability. Here, we used in silico methods to design several constrained peptides comprising a scorpion toxin scaffold, a MDM2 binding epitope, and a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service