Skip to Content
Merck

Skip To

C1295

Cuvette washer

Single

Synonym(s):

cuvette cleaning

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
SB.32
UNSPSC Code:
41121813
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


antibody product type

primary antibodies

General description

Cuvette washer is supplied with a rubber stopper that fits most filter flasks and with neoprene gaskets that form seals with the open ends of the cuvettes.
Supplied with a rubber stopper that fits most filter flasks and with neoprene gaskets that form seals with the open ends of the cuvettes.

Application

Cuvette washer has been used for efficient cuvette cleaning.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Z242721C0793C0918
antibody product type

primary antibodies

antibody product type

-

antibody product type

-

antibody product type

-


Still not finding the right product?

Explore all of our products under Cuvette washer

or

Try our Product Selector Tool to narrow your options




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library