Skip to Content
Merck
All Photos(1)

Key Documents

418846

Sigma-Aldrich

4-Fluorobenzoic acid

99%, purified by sublimation

Synonym(s):

p-Fluorobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CO2H
CAS Number:
Molecular Weight:
140.11
Beilstein:
1906922
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

purified by

sublimation

mp

182-184 °C (lit.)

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)c1ccc(F)cc1

InChI

1S/C7H5FO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,(H,9,10)

InChI key

BBYDXOIZLAWGSL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biodegradation studies of 4-fluorobenzoic acid and 4-fluorocinnamic acid: an evaluation of membrane inlet mass spectrometry as an alternative to high performance liquid chromatography and ion chromatography.
Creaser C, et al.
Analytica Chimica Acta, 454(1), 137-145 (2002)
Jan Marik et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 65(2), 199-203 (2006-08-29)
The widely used bifunctional labeling reagent 4-[18F]fluorobenzoic acid ([18F]FBA) and its activated form N-succinimidyl 4-[18F]fluorobenzoate ([18F]SFB) were prepared using a modified Siemens/CTI chemistry process control unit (CPCU) double vessel [18F]FDG module. The products were obtained with a radiochemical yield greater
Julien Andriès et al.
Bioorganic & medicinal chemistry letters, 20(12), 3730-3733 (2010-05-18)
In search of a serotonin 5-HT(7) radiotracer for positron emission tomography, we developed 1-[2-[(2S)-1-(phenylsulfonyl)pyrrolidin-2-yl]ethyl]piperidin-4-yl 4-fluorobenzoate. After labeling in good yield with fluorine-18 via nitro for fluorine exchange, preliminary biological experiments with autoradiographies failed to evidence any specific 5-HT(7) receptor delineation.
Preparation of 4-fluorophenol and 4-fluorobenzoic acid by the Baeyer-Villiger reaction.
Conte L, et al.
Journal of Fluorine Chemistry, 67(1), 41-45 (1994)
J L Sutcliffe-Goulden et al.
Bioorganic & medicinal chemistry letters, 10(14), 1501-1503 (2000-07-29)
A strategy for the solid phase synthesis of [18F]labelled peptides has been developed. The peptides were prepared on solid support and acylated with 4-[18F]fluorobenzoic acid using HATU within 3 min and the labelled peptide was released from the solid support

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service