Unfortunately, the particle size distribution has not been determined for this product.
1.03819
Iron
for analysis reduced, particle size 10 µm EMSURE®
Synonym(s):
Iron
About This Item
Quality Level
product line
EMSURE®
Assay
≥99.5% (cerimetric)
form
solid
autoignition temp.
>100 °C
potency
30000 mg/kg LD50, oral (Rat)
impurities
≤0.1% Substances insoluble in sulfuric acid
pH
7-9 ( in H2O)
bp
3000 °C/1013 hPa
mp
1535 °C
density
7.87 g/cm3 at 20 °C
bulk density
3000‑4000 kg/m3
anion traces
chloride (Cl-): ≤0.002%
sulfide (S2-): ≤0.002%
cation traces
As: ≤0.0002%
Cu: ≤0.002%
Mn: ≤0.002%
Pb: ≤0.002%
Zn: ≤0.002%
storage temp.
2-30°C
SMILES string
[Fe]
InChI
1S/Fe
InChI key
XEEYBQQBJWHFJM-UHFFFAOYSA-N
Related Categories
Analysis Note
Substances insoluble in sulfuric acid: ≤ 0.1 %
Chloride (Cl): ≤ 0.002 %
Sulphide (S): ≤ 0.002 %
As (Arsenic): ≤ 0.0002 %
Pb (Lead): ≤ 0.002 %
Cu (Copper): ≤ 0.002 %
Mn (Manganese): ≤ 0.002 %
Zn (Zinc): ≤ 0.002 %
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 2 - Self-heat. 2
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
nwg
Flash Point(F)
69.8 °F
Flash Point(C)
21 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Hello, What is the particle size distribution of this iron powder? I expect not all particles to be exactly 10µm in size. Thank you in advance!
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service