Skip to Content
Merck
All Photos(2)

Key Documents

406562

Sigma-Aldrich

Di(propylene glycol) methyl ether acetate, mixture of isomers

99%

Synonym(s):

Dipropylene glycol methyl ether acetate, Dipropylene glycol monomethyl ether acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C3H6OC3H6OCH3
CAS Number:
Molecular Weight:
190.24
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

liquid

refractive index

n20/D 1.417 (lit.)

bp

200 °C (lit.)

density

0.97 g/mL at 25 °C (lit.)

SMILES string

O(C(OC(=O)C)CC)CCCOC

InChI

1S/C9H18O4/c1-4-9(13-8(2)10)12-7-5-6-11-3/h9H,4-7H2,1-3H3

InChI key

USDXBNSCKZJDLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Di(propylene glycol) methyl ether acetate (DPMA, 3-(3-methoxypropoxy)propyl acetate) is extensively employed as a solvent in coatings and silkscreening inks. It is also used as a cleaning agent. In vivo, DPMA rapidly hydrolyzes to dipropylene glycol methyl ether (DPM), thus, the in vivo effects of DPMA are expected to be the same as those of DPM.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dipropylene glycol monomethyl ether: A 13-week inhalation toxicity study in rats and rabbits.
Landry TD and Yano BL.
Toxicological Sciences, 4(4), 612-617 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service