Skip to Content
Merck
All Photos(3)

Key Documents

388378

Sigma-Aldrich

Tributyl O-acetylcitrate

greener alternative

98%

Synonym(s):

Tributyl 2-acetylcitrate, Acetyl tributyl citrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C[CO2(CH2)3CH3][CH2CO2(CH2)3CH3]2
CAS Number:
Molecular Weight:
402.48
Beilstein:
2303316
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.26 psi ( 20 °C)
0.8 psi ( 55 °C)

Assay

98%

form

liquid

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.443 (lit.)

density

1.05 g/mL at 25 °C (lit.)

functional group

ester

greener alternative category

SMILES string

CCCCOC(=O)CC(CC(=O)OCCCC)(OC(C)=O)C(=O)OCCCC

InChI

1S/C20H34O8/c1-5-8-11-25-17(22)14-20(28-16(4)21,19(24)27-13-10-7-3)15-18(23)26-12-9-6-2/h5-15H2,1-4H3

InChI key

QZCLKYGREBVARF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is environmentally-benign and can replace highly toxic phthalates in membrane preparation and thus aligns with "Less Hazardous Chemical Syntheses". Click here for more information.

Application

Tributyl O-acetylcitrate (TBoAC) can be used:
  • As a plasticizer to improve flexibility and impact properties of polylactide (PLA) polymer.
  • As a processing additive for the formulation of bulk heterojunction (BHJ) polymeric organic solar cells (OSCs) to improve their efficiency.
  • In the preparation of semiconducting biopolymer composites of poly(3-hydroxy butyrate) (PHB).
  • In the synthesis of functionalized poly(vinyl chloride)(PVC) membranes for selective separation of perchlorate from water.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

422.6 °F - closed cup

Flash Point(C)

217 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guiding the selection of processing additives for increasing the efficiency of bulk heterojunction polymeric solar cells
Vongsaysy U, et al.
Advanced Energy Materials, 4(3), 1300752-1300752 (2014)
Higher dielectric properties of semiconducting biopolymer composites of poly (3-hydroxy butyrate)(PHB) with polyaniline (PANI), carbon black, and plasticizer
El-hadi AM, et al.
Polymer Bull., 75(4), 1681-1699 (2018)
Mai Bay Stie et al.
Carbohydrate polymers, 242, 116428-116428 (2020-06-23)
Mucoadhesive chitosan-based electrospun nanofibers are promising candidates for overcoming challenges associated with sublingual drug delivery, yet studies focusing on evaluating the mucoadhesive properties of nanofibers for sublingual administration are limited. The aim was to elucidate the mucoadhesive properties of chitosan/polyethylene
Preparation and characterization of functionalized poly (vinyl chloride) membranes for selective separation of perchlorate from water
Wang PY, et al.
Journal of Membrane Science , 476(4), 561-570 (2015)
Panagiota D Zygoura et al.
Journal of chromatography. A, 1093(1-2), 29-35 (2005-10-20)
The determination of commercial plasticizers (di-(2-ethylhexyl)adipate (DEHA) and acetyl tributyl citrate (ATBC)) in aqueous solutions is described. The newly proposed technique of applying microwaves to cloud point extracts in order to enable combination with gas chromatographic analysis has been used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service