382922
Trithiocyanuric acid trisodium salt hydrate
98%
Synonym(s):
1,3,5-Triazine-2,4,6-trithiol trisodium salt hydrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C3N3Na3S3 · xH2O
CAS Number:
Molecular Weight:
243.22 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
powder
mp
82-84 °C (lit.)
SMILES string
O.[Na]Sc1nc(S[Na])nc(S[Na])n1
InChI
1S/C3H3N3S3.3Na.H2O/c7-1-4-2(8)6-3(9)5-1;;;;/h(H3,4,5,6,7,8,9);;;;1H2/q;3*+1;/p-3
InChI key
TZJODYRMPMPAMB-UHFFFAOYSA-K
General description
Trithiocyanuric acid trisodium salt hydrate is the hydrated trisodium salt of trithiocyanuric acid (1,3,5-triazine-2,4,6(1H,3H,5H)-trithione). 2,4,6-trimercapto-s-triazine, trisodium salt commonly referred as TMT-55, is employed for the removal of heavy metals from polluted waters.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Structure and powder diffraction pattern of 2, 4, 6-Trimercapto-s-triazine, trisodium salt (Na3S3C3N3? 9H2O).
Henke KR, et al.
Powder Diffraction, 12(1), 7-12 (1997)
Crystal packing in tetraphenylphosphonium salts of trithiocyanuric acid and its methanol solvate.
Dean PAW, et al.
CrystEngComm, 6(88), 543-548 (2004)
F J Koschier et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 21(4), 495-498 (1983-08-01)
Trithiocyanuric acid (TCY) is a rubber curative of low oral and dermal toxicity and with a low potential for eye and skin irritation. This study determined the effects of TCY on Sprague-Dawley rats at levels of 0.0, 625, 2500 and
Shin Aoki
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 122(12), 1095-1108 (2003-01-04)
We report novel supramolecular polyhedrons formed by three-dimensional self-assembly of multinuclear zinc(II)-12-membered tetraamine (= cyclen = 1,4,7,10-tetraazacyclododecane) complexes with potentially multidentate ligands such as cyanuric acid (CA) and trithiocyanuric acid (TCA) in aqueous solution. Two new supermolecular frameworks were isolated
Mohd Zulkefeli et al.
Inorganic chemistry, 48(19), 9567-9578 (2009-09-12)
A new supramolecular complex, {(Zn(4)L(4))(3)-(TCA(3-))(4)}(12+), was designed and synthesized by the 3:4 self-assembly of a linear tetrakis(Zn(2+)-cyclen) complex (Zn(4)L(4))(8+) and trianionic trithiocyanurate (TCA(3-)) in aqueous solution (cyclen = 1,4,7,10-tetraazacyclododecane). The {(Zn(4)L(4))(3)-(TCA(3-))(4)}(12+) complex, which should have a trigonal prism configuration, was
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service