D129003
1,3,4-Thiadiazole-2,5-dithiol
98%
Synonym(s):
Bismuthiol I, 1,3,4-Thiadiazole-2,5-dithiol, Bismuththiol I
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C2H2N2S3
CAS Number:
Molecular Weight:
150.25
Beilstein:
113444
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
98%
form
powder
mp
162 °C (dec.) (lit.)
SMILES string
Sc1nnc(S)s1
InChI
1S/C2H2N2S3/c5-1-3-4-2(6)7-1/h(H,3,5)(H,4,6)
InChI key
BIGYLAKFCGVRAN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Reagent for the determination of bismuth, copper and lead.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yingchun Fu et al.
Analytical chemistry, 83(17), 6511-6517 (2011-07-26)
We report on the exploitation of metal-organic coordination polymers (MOCPs) as new and efficient matrixes to immobilize enzymes for amperometric biosensing of glucose or phenols. A ligand, 2,5-dimercapto-1,3,4-thiadiazole (DMcT), two metallic salts, NaAuCl(4) and Na(2)PtCl(6), and two enzymes, glucose oxidase
N N Ugarova et al.
Biokhimiia (Moscow, Russia), 45(12), 2243-2253 (1980-12-01)
The individual and combined oxidation of 5-mercapto-1,3,4-thiadiazolthione-2 (bismuthol I) and 3,3'-dimethoxybenzidine (o-dianisidine) by hydrogen peroxide catalyzed by horseradish peroxidase (pH 5,0) was studied. It was shown that bismuthol I is a substrate for peroxidase, which is competitive towards o-dianisidine. In
N Vasimalai et al.
Journal of hazardous materials, 213-214, 193-199 (2012-02-22)
This manuscript describes a highly selective and ultra sensitive determination of Hg(II) in aqueous solution using functionalized mercaptothiadiazole capped silver nanoparticles (AgNPs) by spectrofluorimetry. We have synthesized 2,5-dimercapto-1,3,4-thiadiazole (DMT), 2-mercapto-5-methyl-1,3,4-thiadiazole (MMT) and 2-mercapto-5-amino-1,3,4-thiadiazole (AMT) capped AgNPs by wet chemical method.
Yasuyuki Kiya et al.
Langmuir : the ACS journal of surfaces and colloids, 22(25), 10554-10563 (2006-11-30)
The redox reactions of DMcT at PEDOT-modified glassy carbon electrodes (GCEs) in acetonitrile (AN) have been investigated via cyclic voltammetry (CV) and the electrochemical quartz crystal microbalance (EQCM) in order to elucidate the redox reaction mechanism. A redox couple at
N Vasimalai et al.
The Analyst, 137(14), 3349-3354 (2012-06-12)
The present work describes the determination of picogram Hg(II) using 2,5-dimercapto-1,3,4-thiadiazole stabilized gold nanoparticles (DMT-AuNPs) by a spectrofluorimetry method. DMT-AuNPs show emission maximum at 773 nm with excitation at 514 nm. They show a large stock shift (259 nm), narrow
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service