Skip to Content
Merck
All Photos(1)

Key Documents

336289

Sigma-Aldrich

Titanium boride

powder, <10 μm

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiB2
CAS Number:
Molecular Weight:
69.49
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

form

powder

particle size

<10 μm

density

4.52 g/mL at 25 °C (lit.)

SMILES string

B#[Ti]#B

InChI

1S/2B.Ti

InChI key

QYEXBYZXHDUPRC-UHFFFAOYSA-N

Application

  • Surface Modifications to Reduce Wear in Hot Extrusion of Copper: Discusses the use of titanium boride in surface treatments to enhance the wear resistance of tools used in the hot extrusion of copper (Thewes et al., 2024).
  • Stable DC Vacuum Arc Plasma Generation from a 100 mm TiB2 Cathode: Explores the use of titanium boride in generating stable plasma for coatings, emphasizing its efficiency and stability in industrial applications (Zhirkov et al., 2024).
  • Structure Searches and Superconductor Discovery in XB2: Investigates various titanium boride compounds in the context of their structural properties and potential superconductivity applications (Meng et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G S Vinod Kumar et al.
Physical chemistry chemical physics : PCCP, 9(48), 6415-6425 (2007-12-07)
In the present paper the authors studied isolated metallic films made from the same material used for making metallic foams, and then characterised their properties. Metal films were made from a liquid aluminium alloy reinforced with ceramic particles of known
Benoit Van Aken et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 64(6), 1226-1232 (2012-01-05)
Extracellular DNA in municipal wastewater and effluents from hospitals and R&D laboratories contains antimicrobial resistance and recombinant genes that are today considered as a new class of emerging contaminants. The objective of this research was to investigate the effect of
S Dohshi et al.
Journal of nanoscience and nanotechnology, 1(3), 337-342 (2003-08-14)
Characterizations of Ti-B binary oxide thin films by means of various spectroscopic measurements have shown that Ti-B binary oxide thin films are formed by ultra fine TiO2 nanoparticles. A dramatic decrease in the contact angle of water droplets to 0
Yarui An et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(48), 14439-14446 (2010-11-03)
α-Synuclein (α-SYN) is a very important neuronal protein that is associated with Parkinson's disease. In this paper, we utilized Au-doped TiO(2) nanotube arrays to design a photoelectrochemical immunosensor for the detection of α-SYN. The highly ordered TiO(2) nanotubes were fabricated
Changchun Cao et al.
Huan jing ke xue= Huanjing kexue, 23(6), 108-110 (2003-03-07)
Using 365 nm and 254 nm UV lamp as light source, the degradation of dye X-3B in photolysis, photocatalysis and electrical enhanced photocatalysis was studied in this research. The result shows that the degradation rate of photocatalysis of X-3B was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service