Skip to Content
Merck
All Photos(1)

Key Documents

377414

Sigma-Aldrich

cis-1,3-Dichloropropene

97%

Synonym(s):

(Z )-1,3-Dichloropropene, cis -1,3-Dichloropropylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH=CHCl
CAS Number:
Molecular Weight:
110.97
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

refractive index

n20/D 1.4675 (lit.)

bp

104 °C (lit.)

density

1.225 g/mL at 25 °C (lit.)

SMILES string

ClC\C=C/Cl

InChI

1S/C3H4Cl2/c4-2-1-3-5/h1-2H,3H2/b2-1-

InChI key

UOORRWUZONOOLO-UPHRSURJSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.0 °F - closed cup

Flash Point(C)

23.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Qiuxia Wang et al.
Journal of agricultural and food chemistry, 58(1), 361-365 (2009-11-17)
The gelatin capsule (gel cap) formulation of 1,3-dichloropropene (1,3-D) was developed as a potential means of reducing 1,3-D emissions. The objective of this study was to determine the distribution, emission, and leaching of 1,3-D after applying the gel cap in
Lucrezia Lamastra et al.
Pest management science, 67(11), 1439-1445 (2011-05-14)
1,3-Dichloropropene (1,3-D, CAS No. 542-75-6) is a broad-spectrum soil fumigant used to control numerous species of soilborne plant-parasitic nematodes. 1,3-D consists of two isomers, (Z)- and (E)-1,3-D. There are a number of low-level chlorinated compounds that could potentially be produced
Qiuxia Wang et al.
Journal of agricultural and food chemistry, 57(18), 8414-8420 (2009-09-17)
The gelatin capsule (gel cap) formulation of 1,3-dichloropropene (1,3-D) is a new concept to reduce the environmental release, transport, and hazard potential of the use of 1,3-D to control soilborne diseases and nematodes. The objective of this study is to
Pablo I Nikel et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 368(1616), 20120377-20120377 (2013-03-13)
Chlorinated pollutants are hardly biodegradable under oxic conditions, but they can often be metabolized by anaerobic bacteria through organohalide respiration reactions. In an attempt to identify bottlenecks limiting aerobic catabolism of 1,3-dichloroprop-1-ene (1,3-DCP; a widely used organohalide) in Pseudomonas pavonaceae
S R Yates et al.
Journal of environmental quality, 40(5), 1470-1479 (2011-08-27)
Soil fumigation is important for growing many fruits and vegetable crops, but fumigant emissions may contaminate the atmosphere. A large-scale field experiment was initiated to test the hypothesis that adding composted municipal green waste to the soil surface in an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service