Merck
  • Higher-tier assessment of the potential for groundwater issues due to the use of 1,3-D soil fumigant; evaluation of the active ingredient, metabolites and potentially related chlorinated compounds.
Higher-tier assessment of the potential for groundwater issues due to the use of 1,3-D soil fumigant; evaluation of the active ingredient, metabolites and potentially related chlorinated compounds.
Pest management science (2011-05-14)
Lucrezia Lamastra, Federico Ferrari, Gabriella Fait, Luna Greco, Steve H Kennedy, Ettore Capri, Marco Trevisan
ABSTRACT
1,3-Dichloropropene (1,3-D, CAS No. 542-75-6) is a broad-spectrum soil fumigant used to control numerous species of soilborne plant-parasitic nematodes. 1,3-D consists of two isomers, (Z)- and (E)-1,3-D. There are a number of low-level chlorinated compounds that could potentially be produced as a part of the 1,3-D manufacturing process. 1,3-D and its metabolites, as well as potentially related chlorinated compounds, represent potential groundwater contaminants. This study has investigated the hydrolytic stability, under biotic and abiotic conditions in the laboratory, of an extensive representative list of 1,3-D and potentially related chlorinated compounds in order to predict their environmental fate. All of these compounds showed intrinsic hydrolytic instability under both abiotic and biotic test conditions. Furthermore, a monitoring programme was carried out. Twenty-five wells were monitored in five Italian regions characterised by historical and existing use of 1,3-D. The parent compound, its two major metabolites and potentially related chlorinated compounds were not detected in the well water at > 0.1 µg L(-1) at any sampling location. The soil fumigant (E,Z)-1,3-D, its metabolites and potentially related chlorinated compounds constitute a low risk to groundwater.
Sigma-Aldrich
1,3-Dichloropropene, 90%, technical grade
Supelco
trans-1,3-Dichloropropene, analytical standard
Sigma-Aldrich
cis-1,3-Dichloropropene, 97%