Skip to Content
Merck
All Photos(1)

Key Documents

371890

Sigma-Aldrich

3-Aminopropyl(diethoxy)methylsilane

97%

Synonym(s):

3-(Diethoxymethylsilyl)propylamine, 3-Aminopropyl-methyl-diethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3Si(OC2H5)2(CH2)3NH2
CAS Number:
Molecular Weight:
191.34
Beilstein:
1744264
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.426 (lit.)

bp

85-88 °C/8 mmHg (lit.)

density

0.916 g/mL at 25 °C (lit.)

SMILES string

CCO[Si](C)(CCCN)OCC

InChI

1S/C8H21NO2Si/c1-4-10-12(3,11-5-2)8-6-7-9/h4-9H2,1-3H3

InChI key

HXLAEGYMDGUSBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Amino Surface Modification and Fluorescent Labelling of Porous Hollow Organosilica Particles: Optimization and Characterization.: This study showcases the effectiveness of 3-Aminopropyl(diethoxy)methylsilane in modifying the surface properties of porous hollow organosilica particles, enhancing their application in biosensing and drug delivery systems. The modification process improves particle stability and functionality, demonstrating the compound′s utility in high-performance biomedical applications (Al-Khafaji et al., 2022).
  • Superelastic Multifunctional Aminosilane-Crosslinked Graphene Aerogels for High Thermal Insulation, Three-Component Separation, and Strain/Pressure-Sensing Arrays.: This research highlights the role of 3-Aminopropyl(diethoxy)methylsilane in producing graphene aerogels with exceptional properties such as elasticity, durability, and multi-functionality. These aerogels are promising for thermal insulation, filtration, and as sensors in automotive and aerospace industries, showcasing the compound′s broad potential in advanced material applications (Zu et al., 2019).
  • Improving the Gas-Separation Properties of PVAc-Zeolite 4A Mixed-Matrix Membranes through Nano-Sizing and Silanation of the Zeolite.: Utilizing 3-Aminopropyl(diethoxy)methylsilane for the surface modification of Zeolite 4A, this study enhances the gas-separation efficiency of PVAc-zeolite mixed-matrix membranes. The research addresses significant challenges in membrane-based separation technologies, particularly in the purification processes critical for environmental and industrial applications (Esmaeili et al., 2019).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuexue Xu et al.
Talanta, 205, 120089-120089 (2019-08-28)
We propose a promising capillary immunosensor by combining porous-layer surface modification of an open-tubular capillary and streptavidin-biotin-peroxidase nano-complex signal amplification of chemiluminescence read-out. The immunosensor, namely Porous Layer Open Tubular-Signal Amplification (PLOT-SA) sensor, is successfully applied for ultra-sensitive sensing of
Qishu Qu et al.
Electrophoresis, 37(15-16), 2175-2180 (2016-05-14)
In this work, open-tubular capillary column coated with zeolite imidazolate framework-8 (ZIF-8) nanocrystals was prepared by a layer-by-layer method. The coating was formed by growing ZIF-8 nanocrystals on either bare fused silica capillary wall or the capillary column premodified with
Qihui Zhou et al.
Science advances, 6(5), eaay2756-eaay2756 (2020-02-18)
Artificial rotary molecular motors convert energy into controlled motion and drive a system out of equilibrium with molecular precision. The molecular motion is harnessed to mediate the adsorbed protein layer and then ultimately to direct the fate of human bone
Qishu Qu et al.
Journal of chromatography. A, 1399, 25-31 (2015-05-09)
A simple coating procedure employing a multilayer-by-multilayer process to modify the inner surface of bare fused-silica capillaries with silica nanoparticles was established. The silica nanoparticles were adsorbed onto the capillary wall via a strong electrostatic interaction between amino functional groups
Nazila Esmaeili et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 20(12), 1590-1606 (2019-05-08)
Mixed-matrix membranes containing synthesised nano-sized zeolite 4A and PVAc were fabricated to investigate the effect of zeolite loading on membrane morphology, polymer-filler interaction, thermal stability and gas separation properties. SEM studies revealed that, although the membranes with 40 wt % nano-sized zeolite

Articles

Mesoporous materials, such as aerogels, offer advantages for practical hydrogen storage. They have large surface areas, open porosity, small pore sizes, and the ability to coat the surface with one or more compounds.

Mesoporous materials, such as aerogels, offer advantages for practical hydrogen storage. They have large surface areas, open porosity, small pore sizes, and the ability to coat the surface with one or more compounds.

Mesoporous materials, such as aerogels, offer advantages for practical hydrogen storage. They have large surface areas, open porosity, small pore sizes, and the ability to coat the surface with one or more compounds.

Mesoporous materials, such as aerogels, offer advantages for practical hydrogen storage. They have large surface areas, open porosity, small pore sizes, and the ability to coat the surface with one or more compounds.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service