Skip to Content
Merck
All Photos(2)

Key Documents

104884

Sigma-Aldrich

N-[3-(Trimethoxysilyl)propyl]ethylenediamine

97%

Synonym(s):

[3-(2-Aminoethylamino)propyl]trimethoxysilane, N-(2-Aminoethyl)-3-(trimethoxysilyl)propylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3Si(CH2)3NHCH2CH2NH2
CAS Number:
Molecular Weight:
222.36
Beilstein:
636230
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.444 (lit.)

bp

146 °C/15 mmHg (lit.)

density

1.028 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CO[Si](CCCNCCN)(OC)OC

InChI

1S/C8H22N2O3Si/c1-11-14(12-2,13-3)8-4-6-10-7-5-9/h10H,4-9H2,1-3H3

InChI key

PHQOGHDTIVQXHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-[3-(Trimethoxysilyl)propyl]ethylenediamine has also been used as an organic ligand for the surface modification of silica gel to uptake heavy metal ions. It can react with fluorinated carbon nanotubes (F-CNT) to form aminoalkylalkoxysilane functionalized carbon nanotubes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Sens. 1B - STOT RE 2 Inhalation

Target Organs

respiratory tract irritation

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

276.8 °F - closed cup

Flash Point(C)

136 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adsorption of heavy metal cations by organic ligands grafted on porous materials.
Vasiliev A
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 118(1-3), 251-257 (2009)
Functionalization of single-walled carbon nanotubes with N-[3-(trimethoxysilyl) propyl] ethylenediamine and its cobalt complex.
Oki A
Journal of Physics and Chemistry of Solids, 69(5-6), 1194-1198 (2008)
Adsorption of Cu (II) and Pb (II) onto a grafted silica: isotherms and kinetic models.
Chiron N, at. al.
Water Research, 37(13), 3079-3086 (2003)
T O Acarturk et al.
Journal of biomedical materials research, 44(4), 355-370 (1999-07-09)
Generating skeletal muscle in vitro is an attractive approach to overcome problems associated with autologous transfer of muscle and donor site morbidity during plastic surgery. Such tissue engineering requires application of biomaterials that selectively control the attachment, morphology, and proliferation
Jun-Chao Zuo et al.
Journal of hazardous materials, 235-236, 336-342 (2012-08-25)
Two novel adsorbents to remove excess arsenate and arsenite in the drinking water were prepared for the first time by grafting monoamine and diamine, respectively, and then coordinating Fe(3+) on silica gel that was obtained using sol-gel method with two-step

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service