Skip to Content
Merck
All Photos(2)

Key Documents

368881

Sigma-Aldrich

Fast Red TR Salt

Dye content 15 %

Synonym(s):

Fast Red TR Salt hemi(zinc chloride) salt, 4-Chloro-2-methylbenzenediazonium salt, Azoic Diazo No. 11

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6Cl2N2 · 0.5 ZnCl2
CAS Number:
Molecular Weight:
257.19
Colour Index Number:
37085
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

composition

Dye content, 15%

λmax

284 nm

ε (extinction coefficient)

≥3000 at 281-287 nm in water at 0.006 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].[Cl-].Cl[Zn]Cl.Cc1cc(Cl)ccc1[N+]#N.Cc2cc(Cl)ccc2[N+]#N

InChI

1S/2C7H6ClN2.4ClH.Zn/c2*1-5-4-6(8)2-3-7(5)10-9;;;;;/h2*2-4H,1H3;4*1H;/q2*+1;;;;;+2/p-4

InChI key

RIIVFDKBXXMYRL-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Fast Red TR is a diazonium dye. It serves as a chromogenic substrate for alkaline phosphatase (AP), a common antibody conjugate widely used fin immunohistochemical applications.

Application

Fast Red TR Salt hemi (zinc chloride) salt has been used in the preparation of staining solutions to stain TRACP (tartrate-resistant acid phosphatase). It is also used for the detection of alkaline phosphatase.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An improved in situ hybridization method for the detection of fish pathogens.
McCarthy UM, Urquhart KL, and Bricknell IR
Journal of Fish Diseases, 31(9), 669-677 (2008)
Effect of monoterpenes on the formation and activation of osteoclasts in vitro
Dolder S
Journal of Bone and Mineral Research, 21(4), 647-655 (2006)
Embryonic alkaline phosphatase is expressed at M-phase in the spermatogenic lineage of the mouse
Narisawa S
Development, 116(1), 159-165 (1992)
Kessler C
Nonradioactive Labeling and Detection of Biomolecules (2012)
N Yamaguchi et al.
Applied and environmental microbiology, 62(1), 275-278 (1996-01-01)
An in situ hybridization technique with HNPP (2-hydroxy-3-naphthoic acid-2'-phenylanilide phosphate) and Fast Red TR was used to detect specific bacterial cells at the single-cell level. By this technique, the fluorescent signals of target bacterial cells were up to eight times

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service