Skip to Content
Merck
All Photos(1)

Key Documents

368059

Sigma-Aldrich

Sodium bis(trimethylsilyl)amide solution

0.6 M in toluene

Synonym(s):

Hexamethyldisilazane sodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3Si]2NNa
CAS Number:
Molecular Weight:
183.37
Beilstein:
3629917
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

0.6 M in toluene

density

0.87 g/mL at 25 °C

SMILES string

C[Si](C)(C)N([Na])[Si](C)(C)C

InChI

1S/C6H18NSi2.Na/c1-8(2,3)7-9(4,5)6;/h1-6H3;/q-1;+1

InChI key

WRIKHQLVHPKCJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jerry Isaacson et al.
Angewandte Chemie (International ed. in English), 48(10), 1845-1848 (2009-01-29)
(-)-Dysibetaine has been synthesized in 11 steps from readily available L-malic acid (see scheme). The key step is a unique Ugi 4-center-3-component cyclization reaction, where an ester group acts as the carboxylic acid component. The use of 1,1,1,3,3,3-hexamethyldisilazane as an
A Sebok et al.
Journal of chromatography. A, 1211(1-2), 104-112 (2008-10-14)
This paper presents a derivatization, mass fragmentation study relating to the most common six cholic acids, such as cholic, lithocholic, chenodeoxycholic, ursodeoxycholic, 3-hydroxy,7-ketocholanic and dehydrocholic acids, identified and quantified as pollutants in the aquatic environment at the first time. Derivatizations
Hexamethyldisilazane-mediated controlled polymerization of alpha-amino acid N-carboxyanhydrides.
Hua Lu et al.
Journal of the American Chemical Society, 129(46), 14114-14115 (2007-10-30)
Daniele Fabbri et al.
Environmental science & technology, 42(8), 2957-2963 (2008-05-24)
Using the pyrolysis-gas chromatography-mass spectrometry and off-line pyrolysis/silylation methods for lignites from three Miocene brown coal basins of Poland resulted in the characterization of many organic compounds, including dominant cellulose degradation products such as levoglucosan, 1,6-anhydro-beta-D-glucofuranose, and 1,4:3,6-dianhydroglucopyranose. Levoglucosan is
Marko Marinkovic et al.
Advanced materials (Deerfield Beach, Fla.), 24(29), 4005-4009 (2012-06-26)
A model is presented that describes the gate-voltage-dependent contact resistance and channel-length-dependent charge carrier mobility of small-molecule-based organic thin-film transistors in top and bottom drain/source contact configuration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service