Skip to Content
Merck
All Photos(1)

Key Documents

36580

Sigma-Aldrich

α,α′-Dichloro-m-xylene

≥98.0% (GC)

Synonym(s):

1,3-Bis(chloromethyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH2Cl)2
CAS Number:
Molecular Weight:
175.06
Beilstein:
2079898
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

bp

250-255 °C (lit.)

mp

33-35 °C (lit.)

solubility

methanol: soluble 1 g/10 mL, clear, colorless

density

1.202 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

ClCc1cccc(CCl)c1

InChI

1S/C8H8Cl2/c9-5-7-2-1-3-8(4-7)6-10/h1-4H,5-6H2

InChI key

GRJWOKACBGZOKT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α,α′-Dichloro-m-xylene is an α-chloroxylene derivative and its photolysis in polar and non-polar solvents has been reported. Reactivity of α,α′-dichloro-m-xylene towards [Pt2(μ-S)2(PPh3)4] has been investigated.

Application

α,α′-Dichloro-m-xylene is suitable for use in the synthesis of 1,7-bis(6-methoxy-2-formylphenyl)-1,7-dioxaheptane.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soumen Das et al.
Nuclear medicine and biology, 68-69, 49-57 (2019-02-17)
meta-[123/131I]Iodobenzylguanidine (mIBG) is a clinical agent used for imaging neuroendocrine tumors, where uptake in tumor is via active transport mechanism through norepinephrine transporters (NET). Our group in past have evaluated a 99mTc-analogue of the above tracer, based on 99mTc-4 + 1 labeling
Synthesis and characterization of new macrocyclic schiff bases by the reaction of: 1, 7-Bis (6-methoxy-2-formylphenyl)-1, 7-dioxaheptane and their use in solvent extraction of metals.
Kedy S, et al.
Arabian Journal of Chemistry (2011)
Electrospray mass spectrometric investigation of the reactivity of the sulfide centers in [Pt2 (?-S) 2 (PPh3) 4] towards organic dihalides and the catalytic potential of this complex in the syntheses of organosulfur materials.
Chong SH, et al.
J. Mol. Catal. A: Chem., 204, 267-277 (2003)
Patricia Ponce et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 3(7), 700-705 (2004-07-09)
Photoacid generators (PAG) are chemical systems where light absorption renders strong acid formation, typically with quantum yields greater than one. Many compounds bearing halogen atoms are reported to produce hydrogen halides upon photolysis. Here, alpha-chloroxylene derivatives (ortho, meta and para)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service