Skip to Content
Merck
All Photos(1)

Documents

361461

Sigma-Aldrich

meso-2,3-Butanediol

99%

Synonym(s):

(2R,3S)-2,3-Butanediol, (R,S)-2,3-Butanediol, (erythro-) 2,3-Butanediol, rel-(2R,3S)-2,3-Butanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH(OH)CH3
CAS Number:
Molecular Weight:
90.12
Beilstein:
1718900
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

bp

183-184 °C (lit.)

mp

32-34 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@@H](O)[C@H](C)O

InChI

1S/C4H10O2/c1-3(5)4(2)6/h3-6H,1-2H3/t3-,4+

InChI key

OWBTYPJTUOEWEK-ZXZARUISSA-N

Looking for similar products? Visit Product Comparison Guide

General description

meso-2,3-Butanediol is a diol. 2,3-Butanediol (2,3-DB) exists in three stereoisomeric forms: dextro, levo and meso. 2,3-DB is a crucial chemical feedstock and has wide applications in industry. Production of meso-2,3-butanediol under low oxygen condition by metabolically engineered Escherichia coli is reported. 2,3-Butanediol can be converted to 1,3-butadiene, which is used in the preparation of synthetic rubber. meso-2,3-Butanediol is the source of production of 2-butanol by isolates of lactic acid bacteria.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zheng-Jun Li et al.
Applied microbiology and biotechnology, 87(6), 2001-2009 (2010-05-26)
A metabolically engineered Escherichia coli has been constructed for the production of meso-2,3-butanediol (2,3-BD) under low oxygen condition. Genes responsible for 2,3-BD formation from pyruvate were assembled together to generate a high-copy plasmid pEnBD, in which each gene was transcribed
Payam Ghiaci et al.
FEMS microbiology letters, 360(1), 70-75 (2014-09-02)
2-Butanol has been an issue of industries in many areas, for example, biofuel production (as an advanced alternate fuel), fermented beverages, and food (as taste-altering component). Thus, its source of production, the biological pathway, and the enzymes involved are of
Yuanzhi He et al.
Molecules (Basel, Switzerland), 23(3) (2018-03-23)
(3S)-Acetoin and (2S,3S)-2,3-butanediol are important platform chemicals widely applied in the asymmetric synthesis of valuable chiral chemicals. However, their production by fermentative methods is difficult to perform. This study aimed to develop a whole-cell biocatalysis strategy for the production of
M J Syu
Applied microbiology and biotechnology, 55(1), 10-18 (2001-03-10)
2,3-Butanediol (2,3-BDL), which is very important for a variety of chemical feedstocks and liquid fuels, can be derived from the bioconversion of natural resources. One of its well known applications is the formation of methyl ethyl ketone, by dehydration, which
Jong Myoung Park et al.
Microbial cell factories, 12, 20-20 (2013-02-26)
Klebsiella oxytoca, a Gram-negative, rod-shaped, and facultative anaerobic bacterium, is one of the most promising 2,3-butanediol (2,3-BD) producers. In order to improve the metabolic performance of K. oxytoca as an efficient biofactory, it is necessary to assess its metabolic characteristics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service