Skip to Content
Merck
All Photos(2)

Key Documents

358851

Sigma-Aldrich

Ethyl dithioacetate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CSSCH2CH3
CAS Number:
Molecular Weight:
120.24
Beilstein:
1737711
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.568 (lit.)

bp

61 °C/23 mmHg (lit.)

density

1.048 g/mL at 25 °C (lit.)

functional group

thioether

SMILES string

CCSC(C)=S

InChI

1S/C4H8S2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

KCOPWUJJPSTRIZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl dithioacetate is an alkyl dithioester. It reacts with 5,6-diamino-1,3-dimethyluracil to yield the purine derivative. Raman spectra of ethyl dithioacetate has been recorded as a function of temperature in the liquid phase and in the solid phase at low temperature.

Application

Ethyl dithioacetate may be used in the preparation of N-thioacetylamino acids.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thioacetylation method of protein sequencing: Gas chromatography/Ion trap mass spectrometric detection of 5-acetoxy-2-methylthiazoles.
Stolowitz ML, et al.
Journal of Protein Chemistry, 11(4), 360-361 (1992)
2-?-d-ribofuranosylbenzoxazole from 2, 5-anhydro-d-allonoimidate, and 1, 3-dimethyl-8-?-d-ribofuranosylxanthine from 2, 5-anhydro-d-allono-thioimidates and-dithioates.
El Khadem HS and Kawai J.
Carbohydrate Research, 153(2), 271-283 (1986)
Rotational isomers of ethyl dithioacetate and ethyl thionoacetate: A Raman study.
Verma AL, et al.
Journal of Raman Spectroscopy, 11(5), 390-392 (1981)
Renwu Li et al.
Bioorganic & medicinal chemistry, 28(7), 115356-115356 (2020-02-19)
Past few years have seen an active pursuit of the inhibitors for the deacylation catalyzed by the seven human sirtuins (i.e. SIRT1-7) as valuable chemical biological/pharmacological probes of this enzymatic deacylation and lead compounds for developing novel therapeutics for human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service