Skip to Content
Merck
All Photos(2)

Documents

347604

Sigma-Aldrich

(3-Bromopropyl)trimethylammonium bromide

97%

Synonym(s):

3-Bromo-N,N,N-trimethylpropan-1-aminium Bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)3N(CH3)3Br
CAS Number:
Molecular Weight:
261.00
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

210 °C (dec.) (lit.)

SMILES string

[Br-].C[N+](C)(C)CCCBr

InChI

1S/C6H15BrN.BrH/c1-8(2,3)6-4-5-7;/h4-6H2,1-3H3;1H/q+1;/p-1

InChI key

NNZGNZHUGJAKKT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

(3-Bromopropyl)trimethylammonium bromide is a quaternary ammonium salt and has various applications in organic chemistry, including the synthesis of hydrophilic-hydrophobic block copolymers. Additionally, this compound can be used as a surfactant in the synthesis of ultrathin nano scrolls (NSs).

Application

(3-Bromopropyl)trimethylammonium bromide can be used as a reagent in the synthesis of:
  • Octa-cationic imidazolyl Al(III) phthalocyanine bifunctional catalyst CAT 2.
  • Divalent surfactants like N-(3-trimethylammoniumpropyl)hexadecyldimethylammonium dibromide and N-(3-trimethylammoniumpropyl)octadecylammonium dibromide by reacting with surfactant precursor compounds hexadecyldimethylamine (C16NMe2), octadecyldimethylamine (C18NMe2) respectively.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

General synthesis of large-area flexible bi-atomic subnano thin lanthanide oxide nanoscrolls
Wu M, et al.
Nano Energy, 78, 105318-105318 (2020)
Novel hydrophilic-hydrophobic block copolymer based on cardo poly (arylene ether sulfone) s with bis-quaternary ammonium moieties for anion exchange membranes
Dong X, et al.
Journal of Membrane Science, 518, 31-39 (2016)
Synthesis and property of novel anion exchange membrane based on poly (aryl ether sulfone) s bearing piperidinium moieties
Wang F, et al.
Journal of Membrane Science, 591, 117334-117334 (2019)
K Okazaki et al.
Analytical biochemistry, 145(1), 87-90 (1985-02-15)
Reduced lysozyme was alkylated with 3-bromopropyltrimethylammonium bromide to give reduced and S-3-(trimethylated amino)propylated lysozyme. It was soluble in a wide range of pH and is suitable as the protein substrate to determine protease specificity.
Al (III) phthalocyanine catalysts for CO2 addition to epoxides: Fine-tunable selectivity for cyclic carbonates versus polycarbonates
Gonzalez AC, et al.
Journal of Organometallic Chemistry, 950, 121979-121979 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service