Skip to Content
Merck
(3-Bromopropyl)trimethylammonium bromide 97%

(3-Bromopropyl)trimethylammonium bromide

(3-Bromopropyl)trimethylammonium bromide 97%
Linear Formula:
Br(CH2)3N(CH3)3Br
CAS Number:
Molecular Weight:
261.00

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing