Skip to Content
Merck
All Photos(1)

Key Documents

341436

Sigma-Aldrich

Tetramethyl orthosilicate

≥99%

Synonym(s):

Tetramethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si(OCH3)4
CAS Number:
Molecular Weight:
152.22
Beilstein:
1699658
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

vapor density

5.25 (vs air)

Quality Level

vapor pressure

13 hPa ( 20 °C)

Assay

≥99%

form

liquid

refractive index

n20/D 1.368 (lit.)

bp

121-122 °C (lit.)

mp

−4 °C (lit.)

density

1.023 g/mL at 25 °C (lit.)

SMILES string

CO[Si](OC)(OC)OC

InChI

1S/C4H12O4Si/c1-5-9(6-2,7-3)8-4/h1-4H3

InChI key

LFQCEHFDDXELDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetramethyl orthosilicate is a colorless organosilicon compound consisting of four methoxy groups bonded to a silicon atom. On hydrolysis, it produces silicon dioxide and methanol. It has been widely used in catalysis, synthesis of nanomaterials, ceramics, zeolites, and coating materials.

Application

Tetramethyl orthosilicate can be used:
  • As a precursor to synthesize organic-inorganic coating materials by sol-gel processing method.
  • As an efficient reagent for direct amidation of aliphatic and aromatic carboxylic acids with amines and anilines.
  • As a precursor to fabricate SiO2 nanocomposite films by chemical vapor deposition(CVD) method.
  • As a selective catalyst ofC3-methylation of indole.
Used in the sol-gel synthesis of chromium-doped silicates and in the formation of hexagonal mesoporous silica layers.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetramethyl orthosilicate as a sharp-selective catalyst of C3-methylation of indole by supercritical methanol
I.V.Kozhevnikov, et al.
Journal of Supercritical Fluids, 69, 82-90 (2012)
Fabrication and characteristics of SiOx films by plasma chemical vapor deposition of tetramethylorthosilicate
Mu-Rong Yang, et al.
Surface and Coatings Technology, 123, 204-209 (2000)
Preparation of sol-gel hybrid materials from gamma-methacryloxypropyltrimethoxysilane and tetramethyl orthosilicate: study of the hydrolysis and condensation reactions
Amir A, et al.
Colloid and Polymer Science, 289, 1875-1883 (2011)
Tetramethyl Orthosilicate (TMOS) as a Reagent for Direct Amidation of Carboxylic Acids
D. Christopher Braddock, et al.
Organic Letters, 20, 950-953 (2018)
Carmen Carrasquilla et al.
Journal of the American Chemical Society, 134(26), 10998-11005 (2012-06-26)
Structure-switching, fluorescence-signaling DNA and RNA aptamers have been reported as highly versatile molecular recognition elements for biosensor development. While structure-switching DNA aptamers have been utilized for solid-phase sensing, equivalent RNA aptamers have yet to be successfully utilized in solid-phase sensors

Articles

Reactive silicone chemistry: Focus on pure silicon production, polymerizations, and controlled stereochemistry reactions.

Reactive silicone chemistry: Focus on pure silicon production, polymerizations, and controlled stereochemistry reactions.

Reactive silicone chemistry: Focus on pure silicon production, polymerizations, and controlled stereochemistry reactions.

Reactive silicone chemistry: Focus on pure silicon production, polymerizations, and controlled stereochemistry reactions.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service