Skip to Content
Merck
All Photos(1)

Key Documents

341169

Sigma-Aldrich

Zinc phthalocyanine

Dye content 97 %

Synonym(s):

Phthalocyanine zinc salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H16N8Zn
CAS Number:
Molecular Weight:
577.91
Beilstein:
4121852
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

composition

Dye content, 97%

reaction suitability

reagent type: catalyst
core: zinc

λmax

701 nm

Orbital energy

HOMO 5.17 eV (CV)
LUMO 3.78 eV (CV)

OPV Device Performance

ITO/ZnPc:P60 (1:1)/Bphen/Ag

  • Short-circuit current density (Jsc): 11.7 mA/cm2
  • Open-circuit voltage (Voc): 0.54 V
  • Fill Factor (FF): 0.62
  • Power Conversion Efficiency (PCE): 3.9 %

SMILES string

[Zn]1n2c3nc4nc(nc5n1c(nc6nc(nc2c7ccccc37)c8ccccc68)c9ccccc59)c%10ccccc4%10

InChI

1S/C32H16N8.Zn/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;/h1-16H;/q-2;+2

InChI key

PODBBOVVOGJETB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Zinc (II) phthalocyanine (ZnPc) complexes show photosensitizing properties. Phthalocyanines show significant Π−Π interactions and easy formation of H aggregates.

Application

Zinc phthalocyanine finds general applications in industry (catalysts, photoconductors etc) and biomedical (photodynamic therapy, PDT). ZnPc may be used to photooxidise cyclohexane. A study reports functionalization of polymer membrane with ZnPc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junseok Lee et al.
Chemical communications (Cambridge, England), 49(15), 1545-1547 (2013-01-18)
A photoresponsive drug releasing system based on mesoporous silica nanoparticles (MSN) was developed. A model drug was decorated on the surfaces via singlet oxygen sensitive linkers and a singlet oxygen-generating photosensitizer was loaded in the mesopores of the MSN. An
Jiangtao Ren et al.
Biosensors & bioelectronics, 35(1), 401-406 (2012-04-10)
In this work, a simple and label-free fluorescent method via fluorescent indicator displacement (FID) was proposed for enantioselectively determining d-enantiomer of arginine vasopressin (DV) using DV-specific DNA aptamer (V-apt) and one guanidiniophthalocyanine dye (Zn-DIGP). Zn-DIGP that preferentially binds to single-stranded
Zinc phthalocyanine photocatalyzed oxidation of cyclohexene.
Sehlotho N and Nyokong T
J. Mol. Catal. A: Chem., 219(2), 201-207 (2004)
Sean Marrache et al.
Integrative biology : quantitative biosciences from nano to macro, 5(1), 215-223 (2012-07-27)
A therapeutic technology that combines the phototoxic and immune-stimulating ability of photodynamic therapy (PDT) with the widespread effectiveness of the immune system can be very promising to treat metastatic breast cancer. We speculated that the knowledge of molecular mechanisms of
Ye-Liang Wang et al.
Chimia, 66(1-2), 31-37 (2012-05-02)
We report on high-resolution STM measurements with modified probe tips. First, both the rest atoms and adatoms of a Si(111)-7×7 surface are observed simultaneously. The visibility of rest atoms is dependent upon the sample bias voltage (less than -0.7 V)

Articles

Single-walled carbon nanohorns(SWCNHs) - Find manufacturing, material properties, and applications of carbon nanohorns.

While dye sensitization as the basis for color photography has been accepted for a very long time,1 attempts to use this principle for the conversion of solar light to electricity generally had resulted only in very low photocurrents, below 100 nA/cm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service