Merck
HomeElectron MicroscopySingle-Walled Carbon Nanohorn Properties & Applications
Single-Walled Carbon Nanohorn Properties & Applications
Ryota Yuge
Smart Energy Research Laboratories, NEC Corporation, 34 Miyukigaoka, Tsukuba 305-8501, Japan
Introduction
Single-walled carbon nanohorns (SWCNHs), also called carbon nanohorns (CNHs), are a kind of nano-carbon materials such as carbon nanotubes and graphene, which were discovered by Iijima et al in 1998.1 Each SWCNH has a tubular structure made of a single graphene sheet, which is similar to the single-walled carbon nanotube (SWCNT).2 However, SWCNHs differ considerably from SWCNTs in that they have long cone-shaped tips with cone angles of about 20° and large tube diameters of 2 – 5 nm, and thousands of SWCNHs form spherical aggregates with diameters of 80 – 100 nm (Figure 1). The SWCNHs indicate fine intrinsic properties such as high dispersibility,3 high conductivity,4 and large specific surface area.5 Therefore, potential applications of SWCNHs include gas adsorption,6,7 capacitors,8-10 sensing applications,11 catalytic supports,12-14 composite materials,15 and drug delivery system carriers.16-18 Mass-production technology was developed faster due to the implementation of these applications. Recently, large-scale production of SWCNHs with high purity by CO2 laser ablation of graphite without a catalyst has been performed, and a production rate of 1 kg/day has been achieved.19 In this technical spotlight, we describe the large-scale production method, material properties, and applications of SWCNHs.
TEM images of SWCNHs and SWCNH tip model.
Figure 1.TEM images of SWCNHs and SWCNH tip model.
Large-scale production of SWCNHs
SWCNHs are prepared by CO2 laser ablation of a graphite target without a metal catalyst in an inert gas atmosphere at room temperature.1 Figure 2 shows a schematic illustration of a large-scale production system of SWCNHs.19 This system consists of three chambers, i.e., an exchange chamber (graphite target storage chamber), production chamber, and collection chamber. An inert gas such as argon flows in the inside of the manufacturing system with a pressure of about 760 Torr, and this gas is introduced from the bottom of the laser ablation chamber and evacuated from the top of the collection chamber. The graphite target after CO2 laser ablation is replaced automatically with a new target in the exchange chamber. A large graphite target with a diameter of 100 mm and height of 500 mm is used.
The CO2 laser was operated at 3.5 kW in the continuous emission mode. The target was rotated spirally at 2 rpm for 90 min during the laser ablation, which made continuous production possible. During the laser ablation, the obtained SWCNHs were transferred through the transportation pipe to the collection chamber. In the collection chamber, high-purity SWCNHs were deposited at the bottom of the chamber where there was a gate for attaching an SWCNH storage bottle. When the bottle was filled with SWCNHs, it was detached without interrupting the continuous production of SWCNHs. By using this manufacturing system, SWCNHs were obtained continuously at a rate of 100 g/h, and the SWCNH production of 1kg/day can be achieved.
Schematic illustration of three-chamber system for SWCNH production.
Figure 2.Schematic illustration of three-chamber system for SWCNH production.
Material properties of SWCNHs
3-1. As-grown SWCNHs (Prod. No. 804118)
Figure 3a shows an SEM image of as-grown SWCNHs obtained by using manufacturing equipment.  Each spherical structure was approximately uniform. The diameters of the aggregates were estimated to be about 50-150 nm from direct observation of SEM images.
(a) SEM image and (b) particle-size distribution of SWCNHs.
Figure 3.(a) SEM image and (b) particle-size distribution of SWCNHs.
The particle-size distribution of SWCNHs in the ethanol-dispersion solution was evaluated by dynamic light scattering (DLS) measurements (Figure 3b). Most aggregates existed in the range of 70 to 200 nm, which agrees well with the SEM results. Each SWCNH aggregate was easily mono-dispersed in ethanol. This dispersion was stable and lasted for several hours. The SWCNHs can also disperse in other organic solvents more easily than nanocarbon materials such as carbon black and/or carbon nanotubes. The as-grown SWCNHs did not disperse at all in water because they are hydrophobic.
Figure 4  shows the weight-temperature curves in O2 atmosphere obtained by thermogravimetric analysis (TGA). The derivative curves of the weight-decrease to temperature are also shown; the main peak appears at about 620 °C for SWCNHs. The peak indicates that the combustion of SWCNHs originated from dahlia-type and petal-type SWCNHs.19 Therefore, the purity of the SWCNHs become above 90 wt%. The peak at 760 °C was attributed to micrometer-sized graphitic impurities formed intrinsically from the graphite target by CO2 laser ablation.20
TGA and derivative TGA of SWCNHs.
Figure 4.TGA and derivative TGA of SWCNHs.
3-2. SWCNHs with holes (Holey-SWCNHs) (Prod. No. 804126)
Individual SWCNHs of as-grown SWCNHs are closed, which indicates that the adsorption sites of SWCNHs are only on the outer surface. When as-grown SWCNHs are oxidized, the inside spaces of nanohorns become accessible due to opening of the carbon nanohorn sheaths (holey-SWCNHs). Generally, oxidation treatments are carried out by heat treatment in air,21 acid treatment,22 and H2O2 treatment.23 As a result, the hydrophilic oxygen-containing functional groups are formed mainly on holes of holey-SWCNHs. The holey-SWCNHs can be dispersed highly in water and organic solvents. Generally, the holey-SWCNHs prepared by oxidation with hydrogen peroxide solution at 100 °C,23 have the high dispersibility with a lot of carboxyl groups. The holey-SWCNHs were easily mono-dispersed in ethanol, water, and isopropyl alcohol etc. The dispersions of holey-SWCNHs are more stable than that of as-grown SWCNHs.
Figure 5 shows the TGA and derivative TGA curves of holey-SWCNHs. The main peak at 610 °C and sub-peak at 720 °C of derivative TGA indicate the combustion of the holey-SWCNHs and micrometer-sized graphitic impurities, respectively. The combustion temperature of as-grown SWCNHs and holey-SWCNHs are almost the same, which implies that there is hardly any decrease in the crystallinity of each SWCNH. The purity of the holey-SWCNHs becomes above 90 wt%.
TGA and derivative TGA of holey-SWCNHs.
Figure 5.TGA and derivative TGA of holey-SWCNHs.
Figure 6 shows the N2 adsorption isotherms of SWCNHs and holey-SWCNHs at 77K. The adsorbed amount of nitrogen increased with the increasing relative pressure, the shapes of the adsorption isotherms were almost identical. However, the lower pressure adsorption quantity drastically increased for holey-SWCNHs. The specific surface area of SWCNHs and holey-SWCNHs was estimated at 420 and 1320 m2/g, respectively. The value of holey-SWCNHs was three times larger than that of SWCNHs because the inside spaces of nanohorns become accessible due to opening of the nanohorn sheaths. The maximum previously reported value is 1720 m2/g.10
The inside space of holey-SWCNHs can easily incorporate the gases, drug, metal compounds, solvent, and fullerene etc.6-7, 16, 17, 22, 24 The incorporated materials become stable in the inside space of the SWCNH sheath and can be released again.17 The holes on holey-SWCNHs can be closed again by heat treatment in inert gas.25 The constant-temperature tight-binding molecular dynamics simulation showed that the holes on the sidewalls of the SWCNH sheath were difficult to close by thermal annealing whereas those at the tips were easy to close.25
N2 adsorption isotherms of SWCNHs and holey-SWCNHs at 77K.
Figure 6.N2 adsorption isotherms of SWCNHs and holey-SWCNHs at 77K.
Applications
Since SWCNHs and/or holey-SWCNHs have high specific surface areas, high dispersibility, and high conductivity, they are promising materials for various applications. Some representative applications are as follows.
SWCNHs and/or holey-SWCNHs are novel candidates for gas storage of methane and fluorine6,7 because gas molecules can be stored stably in the cylindrical inside space and interstitial sites. In particular, compressed holey-SWCNHs demonstrated a high storage capacity for methane of 160 cm3/cm3 of nanocarbon at 3.5 MPa and 303 K, exceeding the US Department of Energy target of 150 cm3/cm3 for a practical value expected as storage media.6 The experimental results and grand canonical Monte Carlo simulations show the importance of the adsorption in the interstitial sites for the high total adsorption capacity of holey-SWCNHs.
To energy saving, energy storage technology has attracted much attention in the market.  The capacitor with the holey-SWCNH electrode indicated a large capacity,8-10 which depends strongly on the large specific surface area. The holey-SWCNHs with a specific surface area of 1045 m2/g indicated a specific capacitance of 114 F/g in aqueous electrolyte (H2SO4/H2O).8 A hybrid of holey-SWCNHs and SWCNTs succeeded in preparation of capacitor with large capacity and high power density.9 The SWCNHs and/or holey-SWCNHs can suppress the aggregation of the SWCNTs and undo bundles of SWCNTs.
SWCNHs have been used as the support materials of the catalysts of fuel cells.12-14 The application requires uniform nanoparticles adsorbed on the SWCNH surface. The catalyst sizes, for example, platinum supported on SWCNHs, are controlled to be about 2 nm, which is smaller than that supported on the conventional carbon black.12 This is because there are not only large specific surfaces but also a lot of defective sites and/or interstitial sites between SWCNH sheaths. The current density of the fuel cell using the catalyst-supported SWCNH electrode was larger than that using the catalyst-supported carbon black,12 which demonstrated the suitability of the SWCNHs as electrode material for polymer electrolyte fuel cells.
The SWCNHs with high dispersibility, high conductivity, and unique horn structure can form good composite materials. Polyimide (PI)-based composites containing SWCNHs were fabricated using the spark plasma sintering (SPS) process.15 From evaluation of the wear and friction properties, the specific wear rate of the PI-based SWCNH composite with 5% SWCNHs was in the order of 10-8 mm3/Nm, which is two orders of magnitude less than that of PI alone. The value was slightly superior to that of carbon nanotubes (CNTs) and considerably better than that of graphite. Therefore, the SWCNHs can provide the excellent wear reduction ability. The friction of PI-based composite with 10% SWCNHs and CNTs was lowered. The friction coefficient of PI-based SWCNHs composite was less than 0.25, which is similar to that of PI-based CNTs composite. However, it was slightly higher than that of PI-based composite with 10% graphite.
Various kinds of materials can be incorporated inside holey-SWCNHs, and the incorporated materials can be released easily to the outside. Thus, holey-SWCNHs are expected as a promising carrier material for drug delivery system.16,17 Holey-SWCNHs are especially attractive materials for medical applications since low cytotoxicity has been expected for non-metal catalysts and spherical aggregates with a diameter of 100 nm. The well-known anticancer agent cisplatin (CDDP) can be incorporated in holey-SWCNHs and released from holey-SWCNHs in phosphate buffered saline (PBS).17 As a result, the released CDDP terminated the growth of human lung cancer cells in vitro.
Holey-SWCNHs are also expected as candidate materials for the photodynamic therapy. The carbonaceous tubules absorb light in the near-infrared region. Therefore, the tubule material can annihilate the tumors by photo hyperthermia (PHT) effect. A double photodynamic therapy (PDT) and PHT cancer phototherapy system using a single laser and zinc phthalocyanine (ZnPc, Prod. No. 341169) and protein bovine serum albumin (BSA)-supported holey-SWCNHs was fabricated.18 In this system, ZnPc was the PDT agent, holey-SWCNH was the PHT agent, and the BSA enhanced biocompatibility. The phototherapy effect was confirmed in vitro and in vivo. When the fabricated samples were injected into tumors and transplanted subcutaneously into mice, the tumors almost disappeared on 670-nm laser irradiation.
Materials
Loading
References
1.
Iijima S, Yudasaka M, Yamada R, Bandow S, Suenaga K, Kokai F, Takahashi K. 1999. Nano-aggregates of single-walled graphitic carbon nano-horns. Chemical Physics Letters. 309(3-4):165-170. https://doi.org/10.1016/s0009-2614(99)00642-9
2.
Iijima S, Ichihashi T. 1993. Single-shell carbon nanotubes of 1-nm diameter. Nature. 363(6430):603-605. https://doi.org/10.1038/363603a0
3.
Yuge R, Yudasaka M, Toyama K, Yamaguchi T, Iijima S, Manako T. 2012. Buffer gas optimization in CO2 laser ablation for structure control of single-wall carbon nanohorn aggregates. Carbon. 50(5):1925-1933. https://doi.org/10.1016/j.carbon.2011.12.043
4.
Harada M, Inagaki T, Bandow S, Iijima S. 2008. Effects of boron-doping and heat-treatment on the electrical resistivity of carbon nanohorn-aggregates. Carbon. 46(5):766-772. https://doi.org/10.1016/j.carbon.2008.02.001
5.
Utsumi S, Miyawaki J, Tanaka H, Hattori Y, Itoi T, Ichikuni N, Kanoh H, Yudasaka M, Iijima S, Kaneko K. 2005. Opening Mechanism of Internal Nanoporosity of Single-Wall Carbon Nanohorn. J. Phys. Chem. B. 109(30):14319-14324. https://doi.org/10.1021/jp0512661
6.
Bekyarova E, Murata K, Yudasaka M, Kasuya D, Iijima S, Tanaka H, Kahoh H, Kaneko K. 2003. Single-Wall Nanostructured Carbon for Methane Storage. J. Phys. Chem. B. 107(20):4681-4684. https://doi.org/10.1021/jp0278263
7.
Hattori Y, Kanoh H, Okino F, Touhara H, Kasuya D, Yudasaka M, Iijima S, Kaneko K. 2004. Direct Thermal Fluorination of Single Wall Carbon Nanohorns. J. Phys. Chem. B. 108(28):9614-9618. https://doi.org/10.1021/jp037912i
8.
Yang C, Kim Y, Endo M, Kanoh H, Yudasaka M, Iijima S, Kaneko K. 2007. Nanowindow-Regulated Specific Capacitance of Supercapacitor Electrodes of Single-Wall Carbon Nanohorns. J. Am. Chem. Soc.. 129(1):20-21. https://doi.org/10.1021/ja065501k
9.
Izadi-Najafabadi A, Yamada T, Futaba DN, Yudasaka M, Takagi H, Hatori H, Iijima S, Hata K. 2011. High-Power Supercapacitor Electrodes from Single-Walled Carbon Nanohorn/Nanotube Composite. ACS Nano. 5(2):811-819. https://doi.org/10.1021/nn1017457
10.
Yuge R, Manako T, Nakahara K, Yasui M, Iwasa S, Yoshitake T. 2012. The production of an electrochemical capacitor electrode using holey single-wall carbon nanohorns with high specific surface area. Carbon. 50(15):5569-5573. https://doi.org/10.1016/j.carbon.2012.08.005
11.
Zhang J, Lei J, Xu C, Ding L, Ju H. 2010. Carbon Nanohorn Sensitized Electrochemical Immunosensor for Rapid Detection of Microcystin-LR. Anal. Chem.. 82(3):1117-1122. https://doi.org/10.1021/ac902914r
12.
Yoshitake T, Shimakawa Y, Kuroshima S, Kimura H, Ichihashi T, Kubo Y, Kasuya D, Takahashi K, Kokai F, Yudasaka M, et al. 2002. Preparation of fine platinum catalyst supported on single-wall carbon nanohorns for fuel cell application. Physica B: Condensed Matter. 323(1-4):124-126. https://doi.org/10.1016/s0921-4526(02)00871-2
13.
Kosaka M, Kuroshima S, Kobayashi K, Sekino S, Ichihashi T, Nakamura S, Yoshitake T, Kubo Y. 2009. Single-Wall Carbon Nanohorns Supporting Pt Catalyst in Direct Methanol Fuel Cells. J. Phys. Chem. C. 113(20):8660-8667. https://doi.org/10.1021/jp8105293
14.
Zhang L, Zheng N, Gao A, Zhu C, Wang Z, Wang Y, Shi Z, Liu Y. 2012. A robust fuel cell cathode catalyst assembled with nitrogen-doped carbon nanohorn and platinum nanoclusters. Journal of Power Sources. 220449-454. https://doi.org/10.1016/j.jpowsour.2012.08.009
15.
Tanaka A, Umeda K, Yudasaka M, Suzuki M, Ohana T, Yumura M, Iijima S. 2005. Friction and wear of carbon nanohorn-containing polyimide composites. Tribol Lett. 19(2):135-142. https://doi.org/10.1007/s11249-005-5094-3
16.
Murakami T, Ajima K, Miyawaki J, Yudasaka M, Iijima S, Shiba K. 2004. Drug-Loaded Carbon Nanohorns:  Adsorption and Release of Dexamethasone in Vitro. Mol. Pharmaceutics. 1(6):399-405. https://doi.org/10.1021/mp049928e
17.
Ajima K, Yudasaka M, Murakami T, Maigné A, Shiba K, Iijima S. 2005. Carbon Nanohorns as Anticancer Drug Carriers. Mol. Pharmaceutics. 2(6):475-480. https://doi.org/10.1021/mp0500566
18.
Zhang M, Murakami T, Ajima K, Tsuchida K, Sandanayaka ASD, Ito O, Iijima S, Yudasaka M. 2008. Fabrication of ZnPc/protein nanohorns for double photodynamic and hyperthermic cancer phototherapy. Proceedings of the National Academy of Sciences. 105(39):14773-14778. https://doi.org/10.1073/pnas.0801349105
19.
Azami T, Kasuya D, Yuge R, Yudasaka M, Iijima S, Yoshitake T, Kubo Y. 2008. Large-Scale Production of Single-Wall Carbon Nanohorns with High Purity. J. Phys. Chem. C. 112(5):1330-1334. https://doi.org/10.1021/jp076365o
20.
Fan J, Yudasaka M, Kasuya D, Azami T, Yuge R, Imai H, Kubo Y, Iijima S. 2005. Micrometer-Sized Graphitic Balls Produced Together with Single-Wall Carbon Nanohorns. J. Phys. Chem. B. 109(21):10756-10759. https://doi.org/10.1021/jp050548y
21.
Fan J, Yudasaka M, Miyawaki J, Ajima K, Murata K, Iijima S. 2006. Control of Hole Opening in Single-Wall Carbon Nanotubes and Single-Wall Carbon Nanohorns Using Oxygen. J. Phys. Chem. B. 110(4):1587-1591. https://doi.org/10.1021/jp0538870
22.
Yuge R, Ichihashi T, Shimakawa Y, Kubo Y, Yudasaka M, Iijima S. 2004. Preferential Deposition of Pt Nanoparticles Inside Single-Walled Carbon Nanohorns. Adv. Mater.. 16(16):1420-1423. https://doi.org/10.1002/adma.200400130
23.
Zhang M, Yudasaka M, Ajima K, Miyawaki J, Iijima S. 2007. Light-Assisted Oxidation of Single-Wall Carbon Nanohorns for Abundant Creation of Oxygenated Groups That Enable Chemical Modifications with Proteins To Enhance Biocompatibility. ACS Nano. 1(4):265-272. https://doi.org/10.1021/nn700130f
24.
Miyawaki J, Yudasaka M, Yuge R, Iijima S. 2007. Organic-Vapor-Induced Repeatable Entrance and Exit of C60into/from Single-Wall Carbon Nanohorns at Room Temperature. J. Phys. Chem. C. 111(27):9719-9722. https://doi.org/10.1021/jp069002h
25.
Miyawaki J, Yuge R, Kawai T, Yudasaka M, Iijima S. 2007. Evidence of Thermal Closing of Atomic-Vacancy Holes in Single-Wall Carbon Nanohorns. J. Phys. Chem. C. 111(4):1553-1555. https://doi.org/10.1021/jp067283n