Skip to Content
Merck
All Photos(3)

Documents

335061

Sigma-Aldrich

Ferrocenemethanol

97%

Synonym(s):

FcMeOH, (Hydroxymethyl)ferrocene, Ferrocenylcarbinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12FeO
CAS Number:
Molecular Weight:
216.06
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

reaction suitability

core: iron
reagent type: catalyst

mp

79-81 °C (lit.)

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.OC[C]2[CH][CH][CH][CH]2

InChI

1S/C6H7O.C5H5.Fe/c7-5-6-3-1-2-4-6;1-2-4-5-3-1;/h1-4,7H,5H2;1-5H;

InChI key

QETISSZVCNFQBN-UHFFFAOYSA-N

Application

  • Ferrocenemethanol (FcMeOH) is commonly used as a water soluble ferrocene based reference redox system in electrochemical studies.
  • It is used in the synthesis of ferrocenylmethoxy-isatins, potent antiproliferative agents.
  • FcMeOH can be employed in the preparation of ferrocene-carbohydrate conjugates for biosensing applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jingyi Wang et al.
Journal of colloid and interface science, 570, 350-361 (2020-03-18)
Molybdenite (MoS2) has attracted great attention in the field of advanced materials. MoS2 is normally separated from other minerals by selective flotation, and chemical reagents (so-called depressants) are required to lower the surface hydrophobicity of MoS2. Previous studies had controversial
Effect of the mediator in feedback mode-based SECM interrogation of indium tin-oxide and boron-doped diamond electrodes.
Neufeld A K and O?Mullane A P
Journal of Solid State Electrochemistry, 10(10), 808-816 (2006)
Graphene?based optically transparent electrodes for spectroelectrochemistry in the UV?Vis region.
Weber C M, et al.
Small, 6(2), 184-189 (2010)
Sabine Kuss et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(23), 9249-9254 (2013-05-21)
The emergence of resistance to multiple unrelated chemotherapeutic drugs impedes the treatment of several cancers. Although the involvement of ATP-binding cassette transporters has long been known, there is no in situ method capable of tracking this transporter-related resistance at the
Convenient methods for the synthesis of ferrocene? carbohydrate conjugates.
Casas-Solvas J M, et al.
Organic Letters, 6(21), 3687-3690 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service