Skip to Content
Merck
All Photos(1)

Key Documents

442151

Sigma-Aldrich

Indium(III) trifluoromethanesulfonate

Synonym(s):

In(OTf)3, In(TFA)3, Indium(III) triflate, Trifluoromethanesulfonic acid indium(III) salt, Tris(trifluoromethanesulfonato)indium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO3)3In
CAS Number:
Molecular Weight:
562.03
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: indium
reagent type: catalyst

SMILES string

[In+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/3CHF3O3S.In/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3

InChI key

UCYRAEIHXSVXPV-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Reactant or reagent involved in:
  • Synthesis of stable indium bacteriochlorins
  • Studying basicities of phosphoryl compounds toward triflates Lewis acids
  • Preparation of decahydroquinoline-type toxins via intramolecular hetero Diels-Alder reactions

Catalyst for synthesis of benzoxazoles via cyclocondensations
Reagent used a Lewis acid catalyst in organic synthesis and as a co-catalyst in catalysis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yadav, J. S.; Reddy, B. V. S. et al.
Tetrahedron Letters, 43, 1565-1565 (2002)
Teruhisa Tsuchimoto et al.
Chemical communications (Cambridge, England), (7), 852-853 (2003-05-13)
A palladium-indium triflate catalyst was found to be much more active for the dimerization of vinylarenes compared with generally used cationic palladium(II) catalysts.
Trost, B. M.; Portnoy, M.; Kurihara, H.
Journal of the American Chemical Society, 118, 836-836 (1997)
Frost, C. G.; Chauhan, K. K. et al.
Synlett, 1743-1743 (1999)
Hongliang Wang et al.
ChemSusChem, 11(1), 285-291 (2017-11-15)
Super Lewis acids containing the triflate anion [e.g., Hf(OTf)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service