Skip to Content
Merck
All Photos(2)

Key Documents

333239

Sigma-Aldrich

Titanium(IV) fluoride

Synonym(s):

Titanium tetrafluoride, Titanium fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiF4
CAS Number:
Molecular Weight:
123.86
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder and chunks

reaction suitability

core: titanium
reagent type: catalyst

density

2.798 g/mL at 25 °C (lit.)

SMILES string

F[Ti](F)(F)F

InChI

1S/4FH.Ti/h4*1H;/q;;;;+4/p-4

InChI key

XROWMBWRMNHXMF-UHFFFAOYSA-J

Application

Catalyst for:
  • Chemoselective synthesis and deprotection of geminal diacetates of aldehydes
Used for preparation of glycosyl fluorides; glycosylation; preparation of fluorohydrins; addition of carbanions to aldehydes and imines.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shimizu, M.; Morita, A.; Kaga, T.
Tetrahedron Letters, 40, 8401-8401 (1999)
Carl Hjortsjö et al.
Archives of oral biology, 55(3), 229-234 (2010-02-02)
The protective effect of fluoride solutions against acidic challenge on enamel was tested in vitro using calcium- and profilometer analyses. HF-, SnF(2)-, TiF(4)- and NaF solutions (0.1M F(-), with a pH of 2.0, 2.9, 1.5, 7.3, respectively) and H(2)O were
A Wiegand et al.
Caries research, 43(4), 269-277 (2009-05-15)
This study aimed to analyse and compare the protective effect of buffered (pH 3.5) and native (pH 1.2) TiF(4) in comparison with NaF solutions on enamel erosion. Bovine samples were pretreated with 1.50% TiF(4) or 2.02% NaF (both 0.48 M
Wessel, H. P.
Tetrahedron Letters, 31, 6863-6863 (1990)
Miethchen, R.; Hager, C.; Hein, M.
Synthesis, 159-159 (1997)

Articles

Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.

Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.

Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.

Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service