Skip to Content
Merck
All Photos(1)

Key Documents

324116

Sigma-Aldrich

Trimethylphosphine solution

1.0 M in toluene

Synonym(s):

PMe3, Trimethylphosphorus

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3P
CAS Number:
Molecular Weight:
76.08
Beilstein:
969138
MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

reagent type: ligand
reaction type: Mitsunobu Reaction

reagent type: ligand
reaction type: Wittig Reaction

concentration

1.0 M in toluene

density

0.854 g/mL at 25 °C

functional group

phosphine

SMILES string

CP(C)C

InChI

1S/C3H9P/c1-4(2)3/h1-3H3

InChI key

YWWDBCBWQNCYNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent used in Mitsunobu reaction; transformation of azides into carbamates, aziridines from azidoalcohols; iminophosphoranes and aza-Wittig reaction.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F

Flash Point(C)

-20 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathalie Merceron et al.
Chemical communications (Cambridge, England), (19), 2250-2251 (2002-10-26)
C-Phosphanyl-C-chloroiminium salts formally react as phosphonio(amino)carbenes with tert-butyl isocyanide and trimethylphosphine, and as R2NC+ with vinyl ether and diisopropylamine.
Hughes, D. L.; Reamer, R. A. et al.
Journal of the American Chemical Society, 110, 6487-6487 (110)
Alessandro A Boezio et al.
The Journal of organic chemistry, 68(8), 3241-3245 (2003-04-12)
A new class of orthoacylimine-derived chiral auxiliaries has been synthesized and tested in the nucleophilic addition of organolithium reagents to imines. The precursors can be prepared by an aza-Wittig reaction between the corresponding orthoacyl azide and a variety of aldehydes
A Bondon et al.
Biochimica et biophysica acta, 872(1-2), 163-166 (1986-07-25)
A 31P-NMR study of trimethyl phosphine bound to the heme iron(II) atom in natural myoglobins, hemoglobins and synthetic porphyrin iron(II) shows that the iron-bound trimethylphosphine 31P chemical shifts are sensitive to the presence of globin:separated NMR signals can be observed
N C Goomer et al.
International journal of radiation applications and instrumentation. Part B, Nuclear medicine and biology, 19(6), 679-684 (1992-08-01)
Tc-99m monocationic mixed ligand complexes of phenyl substituted/condensed Schiff's bases, N,N'-ethylene-bis-(benzoylacetone imine) (Lb) or N,N'-ethylene-bis-(salicylaldehyde imine) (Lc) or N,N'-ethylene-bis-(2-hydroxyacetophenone imine) (Ld) and trimethylphosphine were synthesized to determine the influence of the presence of a phenyl group in these tracers on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service