Skip to Content
Merck
All Photos(3)

Key Documents

318116

Sigma-Aldrich

Diethyldithiocarbamic acid diethylammonium salt

97%

Synonym(s):

Diethylammonium diethyldithiocarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NCS2[H2N(C2H5)2]
CAS Number:
Molecular Weight:
222.41
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

82-84 °C (lit.)

functional group

amine

SMILES string

CCNCC.CCN(CC)C(S)=S

InChI

1S/C5H11NS2.C4H11N/c1-3-6(4-2)5(7)8;1-3-5-4-2/h3-4H2,1-2H3,(H,7,8);5H,3-4H2,1-2H3

InChI key

NBGTWXBPCIHUQD-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shailaja P Rao et al.
PloS one, 8(5), e65155-e65155 (2013-06-07)
Previously we identified palmitoyl-, oleoyl-, linoleoyl-, and arachidonoyl-lysophosphatidylcholine (LPC 16:0, 18:1, 18:2 and 20:4) as the most prominent LPC species generated by endothelial lipase (EL). In the present study, we examined the impact of those LPC on acetylcholine (ACh)- induced
Chulun Shao et al.
ACS nano, 12(12), 12629-12637 (2018-11-30)
The specific characteristics of the tumor vascular microenvironment such as microvascular permeability and water diffusion have been demonstrated to play essential roles in the evaluation of infiltration of tumors. However, at present, there are few contrast agents (CAs) for magnetic
Muhammad Sajid et al.
Molecules (Basel, Switzerland), 25(15) (2020-07-31)
In this work, we developed an extraction technique that can handle simple as well as complex matrixed liquid (aqueous) samples. In the standard liquid-liquid extraction, it is quite challenging to deal with complex liquid samples as they may complicate the
Qian Wang et al.
Neuropeptides, 80, 102021-102021 (2020-02-09)
Neuropeptides are involved in the regulation of the sympathetic activity and blood pressure in the paraventricular nucleus of the hypothalamus (PVN). The present study was designed to determine how alarin modulates the renal sympathetic nerve activity (RSNA), arterial blood pressure
Juan J Pinto et al.
Talanta, 142, 84-89 (2015-05-25)
During the last decade, hollow fiber liquid phase micro-extraction (HF-LPME) has become an attractive alternative in sample treatment for the analysis of trace metals in seawater. If compared with other similar methodologies, its main advantages are associated to a higher

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service