Skip to Content
Merck
All Photos(1)

Key Documents

359548

Sigma-Aldrich

Ammonium diethyldithiocarbamate

Synonym(s):

Diethyldithiocarbamic acid ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NC(S)SSNH4
CAS Number:
Molecular Weight:
166.31
Beilstein:
5796362
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

80 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

N.CCN(CC)C(S)=S

InChI

1S/C5H11NS2.H3N/c1-3-6(4-2)5(7)8;/h3-4H2,1-2H3,(H,7,8);1H3

InChI key

JSYCNFYVINPILE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium diethyldithiocarbamate is suitable for use in a study to detect vascular O2- production in intact aortic segments by lucigenin-enhanced chemiluminescence method.
Reactant involved in:
  • Reactions with copper acetylacetonato complex
  • Triazole compounds
  • Causing accelerated degradation of methyl iodide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Gómez-Roso et al.
British journal of pharmacology, 158(8), 1911-1921 (2009-11-18)
The aim of this study was to investigate the effect of chronic treatment with antihypertensive and non-antihypertensive doses of zofenopril on cardiovascular changes in spontaneously hypertensive rats (SHR). Male SHR were treated with 0.5 or 10 mg kg(-1) per day
Solubility of Diethyl Ammonium Diethyldithiocarbamate (Et~ 2NH~ 2DDC) in Supercritical Carbon Dioxide.
Wang H-b, et al.
Journal of Chemical Engineering of Chinese Universities / Gao Xiao Hua Xue Gong Cheng Xue Bao, 15(5), 472-475 (2001)
J Llorente et al.
The Journal of pharmacology and experimental therapeutics, 342(2), 552-560 (2012-05-18)
It has previously been shown that nitric oxide (NO) synthase is involved in the development of opioid tolerance. The aim of the present work was to study the effect of NO on μ-opioid receptor (MOR) desensitization. Furthermore, we explored the
Xueqin Guo et al.
Talanta, 94, 70-76 (2012-05-23)
A new method of solidified floating organic drop microextraction (SFODME) combined with electrothermal vaporization (ETV)-inductively coupled plasma mass spectrometry (ICP-MS) was developed for the determination of trace heavy metals in environmental water samples with sodium diethyldithiocarbamate (DDTC) as both chelating
Kuno Lehmann et al.
Annals of surgery, 256(5), 730-737 (2012-10-26)
The aim of hyperthermic intraperitoneal chemotherapy (HIPEC) is to eradicate microscopic residual tumor after radical surgical tumor excision in patients with peritoneal carcinomatosis. The common use of antineoplastic agents such as mitomycin C, doxorubicin, or oxaliplatin with hyperthermia fails to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service