Skip to Content
Merck
All Photos(1)

Key Documents

317519

Sigma-Aldrich

Methyl 2-acetamidoacrylate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(NHCOCH3)CO2CH3
CAS Number:
Molecular Weight:
143.14
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

bp

104 °C/8 mmHg (lit.)

mp

50-52 °C (lit.)

SMILES string

COC(=O)C(=C)NC(C)=O

InChI

1S/C6H9NO3/c1-4(6(9)10-3)7-5(2)8/h1H2,2-3H3,(H,7,8)

InChI key

SMWNFFKPVLVOQQ-UHFFFAOYSA-N

General description

Conjugated addition of secondary amines, imidazole and pyrazole to methyl 2 methyl 2-acetamidoacrylate in the presence of a catalyst results in the formation of β-Dialkylamino-α-alanine and β-(N-heteroaryl)-α-alanine derivatives. Methyl-2-acetamidoacrylate (M2AA) is an anti-inflammatory agent. The catalytic reaction of methyl 2-acetamidoacrylate with Grignard′s reagents affords α-amino esters. M2AA can form thermosensitive copolymers with methyl acrylate.
Methyl ester of 2-acetamidoacrylate . methyl 2-acetamidoacrylate (Me-2-AA) is a di-unsaturated α-amino acid derivative. methyl-2-acetamidoacrylate exihibits anti -inflammatory properties, it is very effective against lipopolysaccharide (LPS)- induced nitric oxide production by RAW 264.

Application

Methyl 2-acetamidoacrylate can undergo [2+2] cycloaddition (Michael–Dieckmann-type reaction) with ketene diethyl acetal to yield the cyclobutane core. It may be used in rhodium-catalyzed 2-alkenylpyrrole formation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Madeleine E Kieffer et al.
Journal of the American Chemical Society, 134(11), 5131-5137 (2012-03-07)
The tandem Friedel-Crafts conjugate addition/asymmetric protonation reaction between 2-substituted indoles and methyl 2-acetamidoacrylate is reported. The reaction is catalyzed by (R)-3,3'-dibromo-BINOL in the presence of stoichiometric SnCl(4), and is the first example of a tandem conjugate addition/asymmetric protonation reaction using
FeCl3-catalyzed conjugate addition of secondary amines, imidazole and pyrazole to methyl 2-acetamidoacrylate.
Montserrat P and Pleixats R
Tetrahedron, 51(30), 8355-8362 (1995)
Thermosensitive properties of a novel poly(methyl 2-acetamidoacrylate-co-methyl acrylate)
Okamura H, et al.
European Polymer Journal, 38(4), 639-644 (2002)
Ethyl pyruvate: a novel anti?inflammatory agent.
Fink MP, et al.
J. Int. Med., 261(4), 349-362 (2007)
Benoît Cossec et al.
Molecules (Basel, Switzerland), 13(10), 2394-2407 (2008-10-03)
A safe and simple method for methyl S-arylmercapturate synthesis is described. Thirteen such compounds, to be used afterwards in metabolism studies, have been obtained with yields ranging from 71 to 99.6%. These compounds were obtained using a sulfa-Michael addition and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service