Skip to Content
Merck
All Photos(2)

Documents

552348

Sigma-Aldrich

2-Carboxyethyl acrylate

contains 900-1100 ppm MEHQ as inhibitor

Synonym(s):

β-(Acryloyloxy)propionic acid, β-Carboxyethyl acrylate, 3-(Acryloyloxy)propionic acid, 3-Acryloyloxypropanoic acid, Acrylic acid 2-carboxyethyl ester, Acrylic acid dimer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCO2(CH2)2CO2H
CAS Number:
Molecular Weight:
144.13
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

contains

900-1100 ppm MEHQ as inhibitor

refractive index

n20/D 1.457 (lit.)

pH

2.95 (10 wt. % in H2O)

bp

103 °C/19 mmHg (lit.)

solubility

H2O: soluble

density

1.214 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OC(=O)CCOC(=O)C=C

InChI

1S/C6H8O4/c1-2-6(9)10-4-3-5(7)8/h2H,1,3-4H2,(H,7,8)

InChI key

CYUZOYPRAQASLN-UHFFFAOYSA-N

Application

Can be polymerized in solution or emulsion to produce acrylic, vinyl-acrylic, or styrenic-acrylic polymers with greater flexibility due to lower glass transition of itshomopolymer (<30C) and improved adhesion.

Features and Benefits

More reactive in its salt form than acylic acid, allowing high levels of incorporation over awide pH range. Also see Poly(2-carboxyethyl) acrylate, 407585.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of acrylic acid dimer on the properties of carboxycement.
S Pokorný et al.
Journal of dentistry, 16(2), 76-79 (1988-04-01)
Mi-Sook Kim et al.
Journal of biomedical materials research. Part A, 83(3), 674-682 (2007-05-29)
We developed and characterized a novel in situ chitosan-poly(ethylene oxide) (PEO) hydrogel via two steps: 2-carboxyethyl acrylate molecules were grafted to the primary amine functional groups in chitosan in the first step and then Michael type addition reaction was processed
Nadezhda Vorzobova et al.
Polymers, 11(12) (2019-12-11)
The possibility of the application of acrylate compositions and Bayfol HX photopolymers in holographic technologies is considered. The holographic characteristics of materials, their advantages, and limitations in relation to the tasks of obtaining holographic elements based on periodic structures are
Kecheng Li et al.
Macromolecular rapid communications, 41(18), e2000208-e2000208 (2020-06-13)
Although a range of polymer-protein polyion complex (PIC) micelle systems have been developed in the literature, relatively little attention has been paid to the influence of polymer structure on the assembly, or to the mechanism of disassembly. In this work
Rachel Chapla et al.
Polymers, 12(12) (2020-12-20)
Local mechanical stiffness influences cell behavior, and thus cell culture scaffolds should approximate the stiffness of the tissue type from which the cells are derived. In synthetic hydrogels, this has been difficult to achieve for very soft tissues such as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service