Skip to Content
Merck
All Photos(2)

Key Documents

308501

Sigma-Aldrich

Benzyl butyl phthalate

98%

Synonym(s):

Butyl Benzyl Pthalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-[CH3(CH2)3O2C]C6H4CO2CH2C6H5
CAS Number:
Molecular Weight:
312.36
Beilstein:
2062204
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

10.8 (vs air)

Quality Level

vapor pressure

0.16 mmHg ( 150 °C)

Assay

98%

form

viscous liquid

autoignition temp.

450 °F

refractive index

n20/D 1.54 (lit.)

density

1.1 g/mL at 25 °C (lit.)

SMILES string

CCCCOC(=O)c1ccccc1C(=O)OCc2ccccc2

InChI

1S/C19H20O4/c1-2-3-13-22-18(20)16-11-7-8-12-17(16)19(21)23-14-15-9-5-4-6-10-15/h4-12H,2-3,13-14H2,1H3

InChI key

IRIAEXORFWYRCZ-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)

Looking for similar products? Visit Product Comparison Guide

Application

  • Use as Plasticizer: BBP was predominantly used as a plasticizer in PVC to increase flexibility. Its application extended to products such as artificial leather, flooring, and adhesives (Wikiwand).

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asma Verdian et al.
Journal of hazardous materials, 402, 123531-123531 (2020-07-30)
Polychlorinated biphenyls (PCBs) are considered persistent bio-accumulative toxicants which threats global food safety and environmental health. Traditional analytical techniques for detection of PCBs are time-consuming and they do not satisfy urgent need for rapid and accurate monitoring of these persistent
Hongyuan Yan et al.
The Analyst, 135(10), 2585-2590 (2010-08-18)
A simple and rapid sample pretreatment procedure based on ultrasound-assisted dispersive liquid-liquid microextraction is proposed for the determination of four phthalate esters (dibutyl phthalate, butyl benzyl phthalate, diisooctyl phthalate, dioctyl phthalate) in bottled water samples using gas chromatography-flame ionization detection.
Rosario Planelló et al.
Aquatic toxicology (Amsterdam, Netherlands), 105(1-2), 62-70 (2011-06-21)
In this work, the effects of butyl benzyl phthalate (BBP) and di(2-ethylhexyl) phthalate (DEHP), two of the most extensively used phthalates, were studied in Chironomus riparius under acute short-term treatments, to compare their relative toxicities and identify genes sensitive to
Susan Sumner et al.
Journal of applied toxicology : JAT, 29(8), 703-714 (2009-09-05)
This study was conducted to evaluate the use of metabolomics for improving our ability to draw correlations between early life exposures and reproductive and/or developmental outcomes. Pregnant CD rats were exposed by gavage daily during gestation to vehicle or to
Wen-Ching Chen et al.
Ecotoxicology and environmental safety, 74(5), 1325-1330 (2011-04-19)
This study examined the effect of benzyl butyl phthalate (BBP), a phthalate ester (PAE) and an endocrine disruptor, on water celery, Ipomoea aquatica Forsk., one of the most popular leaf vegetables in Taiwan. After 28 days of cultivation, treatment with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service