Skip to Content
Merck
All Photos(3)

Key Documents

307394

Sigma-Aldrich

Guanidine sulfate salt

99%

Synonym(s):

Bisguanidinium sulfate, Guanidinium sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2C(=NH)NH2]2 · H2SO4
CAS Number:
Molecular Weight:
108.11
Beilstein:
3632154
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

290-293 °C (dec.) (lit.)

SMILES string

NC(N)=N.NC(N)=N.OS(O)(=O)=O

InChI

1S/2CH5N3.H2O4S/c2*2-1(3)4;1-5(2,3)4/h2*(H5,2,3,4);(H2,1,2,3,4)

InChI key

UYAKTBUPMOOXNW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The multicomponent condensation of guanidine sulfate salt with H2S and CH2O was studied.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multicomponent reactions of urea and its derivatives with CH2O and H2S in the synthesis of 1, 3, 5-thiadiazinane-4-(thi) ones and macroheterocycles.
ARKIVOC (Gainesville, FL, United States), 8, 149-162 (2011)
Manuel Tzouros et al.
European journal of mass spectrometry (Chichester, England), 19(1), 57-69 (2013-07-12)
A combination of high-performance liquid chromatography (HPLC) and atmospheric-pressure chemical ionization mass spectrometry (APCI-MS and APCI-MS/MS) was used to detect and characterize linear polyamine derivatives in the venom of the spiders Agelenopsis aperta, Hololena curta and Paracoelotes birulai. The compounds
Yuling Shao et al.
Journal of molecular graphics & modelling, 40, 54-63 (2013-01-29)
Density functional theory and volume-based thermodynamics calculations have been performed to study the crystal densities, heats of formation (HOFs), energetic properties, and thermodynamics of formation for a series of ionic salts composed of triaminoguanidinium or ammonium cations and tetrazole-based anions.
Christoph J O Kaiser et al.
Nature communications, 4, 1571-1571 (2013-03-14)
Neurodegeneration is linked to protein aggregation in several human disorders. In Huntington's disease, the length of a polyglutamine stretch in Huntingtin is correlated to neuronal death. Here we utilize a model based on glutamine stretches of 0, 30 or 56
Palani Elumalai et al.
Inorganic chemistry, 52(4), 1883-1894 (2013-02-09)
Reaction of cis-[Cl(2)Pt(S(O)Me(2))(2)] with 1 equiv of sym-N,N',N″-triarylguanidines, ArN═C(NHAr)(2) (sym = symmetrical; Ar = 2-MeC(6)H(4) (LH(2)(2-tolyl)), 2-(MeO)C(6)H(4) (LH(2)(2-anisyl)), 4-MeC(6)H(4) (LH(2)(4-tolyl)), 2,5-Me(2)C(6)H(3) (LH(2)(2,5-xylyl)), and 2,6-Me(2)C(6)H(3) (LH(2)(2,6-xylyl))) in toluene under reflux condition for 3 h afforded cis- or trans-[Cl(2)Pt(S(O)Me(2))(ArN═C(NHAr)(2))] (Ar = 2-MeC(6)H(4)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service