Merck
  • Structure-property relationships of energetic nitrogen-rich salts composed of triaminoguanidinium or ammonium cation and tetrazole-based anions.
Structure-property relationships of energetic nitrogen-rich salts composed of triaminoguanidinium or ammonium cation and tetrazole-based anions.
Journal of molecular graphics & modelling (2013-01-29)
Yuling Shao, Weihua Zhu, Heming Xiao
ABSTRACT
Density functional theory and volume-based thermodynamics calculations have been performed to study the crystal densities, heats of formation (HOFs), energetic properties, and thermodynamics of formation for a series of ionic salts composed of triaminoguanidinium or ammonium cations and tetrazole-based anions. Substitution with --NF₂, --CH₂NF₂, --CF₂NF₂, or --C(NO₂)₂NF₂ groups increased the densities of the salts. The densities of the tetrazole-based salts are affected not only by different substituents but also by different cations. The --CN or --N₃ groups are effective substituents for increasing the HOFs of the salts. The triaminoguanidinium cation is more effective than the ammonium cation for increasing the HOF of the tetrazole-based salts. Substitution with --NO₂, --NF₂, or --C(NO₂)₂NF₂ groups enhances the explosive properties of the salts. The thermodynamics of formation of the salts reveal that all of the tetrazole-based salts with the triaminoguanidinium or ammonium cation could be synthesized using the proposed reactions. Our calculated methods provide a straightforward and inexpensive route for screening a large number of potentially energetic ionic salts.
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Guanidine sulfate salt, 99%
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Guanidine hydrochloride, for molecular biology, ≥99%