Skip to Content
Merck
All Photos(1)

Documents

307009

Sigma-Aldrich

Propyl benzoate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CO2CH2CH2CH3
CAS Number:
Molecular Weight:
164.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.501 (lit.)

bp

229.5 °C/766 mmHg (lit.)

density

1.026 g/mL at 25 °C (lit.)

SMILES string

CCCOC(=O)c1ccccc1

InChI

1S/C10H12O2/c1-2-8-12-10(11)9-6-4-3-5-7-9/h3-7H,2,8H2,1H3

InChI key

UDEWPOVQBGFNGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The adsorption data of propyl benzoate was obtained by frontal analysis on a symmetry-C18 column by a mixture of methanol and water as the mobile phase.

Application

Propyl benzoate was used to extract desipramine, a model compound used in the electrochemical analysis by a combined hollow fiber-based liquid three-phase microextraction and voltammetric method.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Saraji et al.
Talanta, 82(4), 1588-1593 (2010-08-31)
In this paper, a combined hollow fiber-based liquid three-phase microextraction and voltammetric method are applied for the first time as a highly selective and sensitive method of electrochemical analysis. Desipramine, used as a model compound was extracted from 8 mL
Fabrice Gritti et al.
Journal of chromatography. A, 1069(1), 31-42 (2005-04-23)
The adsorption data of propyl benzoate were acquired by frontal analysis (FA) on a Symmetry-C18 column, using a mixture of methanol (65%, v/v) and water as the mobile phase, at three different flow rates, 0.5, 1.0 and 2.0 mL/min. The
Suvi-Katriina Ruokonen et al.
Journal of chromatography. A, 1479, 194-203 (2016-12-14)
Liposomes were used as biomimetic models in capillary electrokinetic chromatography (EKC) for the determination of distribution constants (K
Filip Duša et al.
Electrophoresis, 37(2), 363-371 (2015-10-16)
The use of bare fused silica capillary in CE can sometimes be inconvenient due to undesirable effects including adsorption of sample or instability of the EOF. This can often be avoided by coating the inner surface of the capillary. In
Makiko Fujii et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 109, 280-287 (2017-08-20)
We studied the effect that three alcohols, ethanol (EA), propanol (PA), and isopropanol (IPA), have on the skin permeation of p-hydroxy benzoic acid methyl ester (HBM), a model ester-type prodrug. HBM was applied to Yucatan micropig skin in a saturated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service