Skip to Content
Merck
All Photos(1)

Key Documents

339725

Sigma-Aldrich

Chloromethane solution

1.0 M in tert-butyl methyl ether, anhydrous

Synonym(s):

Methyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3Cl
CAS Number:
Molecular Weight:
50.49
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

anhydrous

form

liquid

concentration

1.0 M in tert-butyl methyl ether

impurities

<0.005% water

density

0.765 g/mL at 25 °C

functional group

alkyl halide
chloro

storage temp.

2-8°C

SMILES string

CCl

InChI

1S/CH3Cl/c1-2/h1H3

InChI key

NEHMKBQYUWJMIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Radiolysis of aqueous solutions of chloromethanes was performed with γ-rays. Dechlorination of chloromethane on iron and palladium-iron bimetallic surface in aqueous systems has been reported.

Packaging

Supplied in a Sure/Pac cylinder and has a 1/4-turn bronze ball valve with a female 1/4" NPT outlet thread (Z122661) installed. Before using the cylinder, ensure that the valve is closed, then remove carbon steel plug that seals the outlet valve.

Compatible with the following:For instructions please refer to technical bulletin AL-136: The Aldrich Sure/Pac Cylinder System

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

septum inlet adapter

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Carc. 2 - Flam. Liq. 2 - Ozone 1 - Repr. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-29.2 °F - closed cup

Flash Point(C)

-34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dechlorination of chloromethanes on iron and palladium-iron bimetallic surface in aqueous systems.
Wan C, et al
Environmental Toxicology and Chemistry / Setac, 18(6), 1091-1096 (1999)
L El Khoury et al.
The Journal of chemical physics, 136(2), 024319-024319 (2012-01-21)
We present a combined experimental and theoretical study of isolated CH(3)Cl molecules using resonant inelastic x-ray scattering (RIXS). The high-resolution spectra allow extraction of information about nuclear dynamics in the core-excited molecule. Polarization-resolved RIXS spectra exhibit linear dichroism in the
Xiaofeng Tang et al.
The Journal of chemical physics, 136(3), 034304-034304 (2012-01-28)
Utilizing threshold photoelectron-photoion coincidence (TPEPICO) velocity imaging, dissociation of state-selected CH(3)Cl(+) ions was investigated in the excitation energy range of 11.0-18.5 eV. TPEPICO time-of-flight mass spectra and three-dimensional time-sliced velocity images of CH(3)(+) dissociated from CH(3)Cl(+)(A(2)A(1) and B(2)E) ions were
High dose radiolysis of aqueous solutions of chloromethanes: Importance in the storage of radioactive organic wastes.
Rajesh P, et al.
Journal of Nuclear Materials, 361(1), 10-17 (2007)
Francesca Bleken et al.
The journal of physical chemistry. A, 114(27), 7391-7397 (2010-06-19)
The methylation of ethene by methyl chloride and methanol in the microporous materials SAPO-34 and SSZ-13 has been studied using different periodic atomistic modeling approaches based on density functional theory. The RPBE functional, which earlier has been used successfully in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service