Skip to Content
Merck
All Photos(1)

Documents

305065

Sigma-Aldrich

1,2,4,5-Benzenetetramine tetrahydrochloride

≥95%

Synonym(s):

1,2,4,5-Tetraaminobenzene tetrahydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H2(NH2)4·4HCl
CAS Number:
Molecular Weight:
284.01
Beilstein:
3701344
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

≥95%
≥95.0% (AT)

mp

≥300 °C (lit.)

SMILES string

Cl.Cl.Cl.Cl.Nc1cc(N)c(N)cc1N

InChI

1S/C6H10N4.4ClH/c7-3-1-4(8)6(10)2-5(3)9;;;;/h1-2H,7-10H2;4*1H

InChI key

BZDGCIJWPWHAOF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2,4,5-Benzenetetraminetetrahydrochloride is widely used as a novel source of carbon and nitrogen insynthesizing benzimidazole-linked polymers and carbon dot nanomaterials.

Application

1,2,4,5-Benzenetetramine tetrahydrochloride can be used as a precursor in the synthesis of:
  • A pyrene-based benzimidazole-linked polymer by the co-condensation with 1,3,6,8-tetrakis(4-formylphenyl)pyrene in dimethylformamide.
  • Triazine-based benzimidazole-linked polymers for highly selective CO2 capture.
  • Nitrogen-doped carbon dots (NCDs) by simple hydrothermal method, applicable as a highly selective fluorescent probe for sensing Mg2+ ions in an aqueous solution.
  • Red-emitting carbon dots via the solvothermal method.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Halszka Ponamarczuk et al.
Frontiers in bioscience (Landmark edition), 23, 1612-1627 (2018-01-03)
Protein disulfide isomerase (PDI) is an abundant reticulum endoplasmic protein but also acts as an important functional regulator of some extracellular surface proteins. Recent studies suggest that PDI plays a role in integrin activation and thrombus formation. The aim of
Ali M Ethaeb et al.
Apoptosis : an international journal on programmed cell death, 25(1-2), 29-41 (2019-10-28)
Accumulation of tissue factor (TF) within cells leads to cellular apoptosis mediated through p38 and p53 pathways. In this study, the involvement of Src1 in the induction of TF-mediated cell apoptosis, and the mechanisms of Src1 activation were investigated. Human
Arun Kumar Nalla et al.
PloS one, 5(9), e13006-e13006 (2010-10-05)
Despite effective radiotherapy for the initial stages of cancer, several studies have reported the recurrence of various cancers, including medulloblastoma. Here, we attempt to capitalize on the radiation-induced aggressive behavior of medulloblastoma cells by comparing the extracellular protease activity and
Yeongseon Byeon et al.
Cancer research, 78(21), 6247-6256 (2018-08-18)
Chemotherapy is commonly used in the treatment of ovarian cancer, yet most ovarian cancers harbor inherent resistance or develop acquired resistance. Therefore, novel therapeutic approaches to overcome chemoresistance are required. In this study, we developed a hyaluronic acid-labeled poly(d,l-lactide-co-glycolide) nanoparticle
Jie Zhou et al.
International journal of molecular medicine, 40(6), 1649-1656 (2017-10-19)
This study aimed to investigate the role of focal adhesion kinase (FAK) signaling in the inhibitory effects of black rice anthocyanins (BRACs) on human epidermal growth factor receptor-2 (HER-2)-positive human breast cancer cell metastasis, using the MCF-10A, MCF-7 and MDA-MB-453 cells. BRACs exerted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service