Skip to Content
Merck
All Photos(4)

Key Documents

N818

Sigma-Aldrich

1,4,5,8-Naphthalenetetracarboxylic dianhydride

Synonym(s):

NTCDA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H4O6
CAS Number:
Molecular Weight:
268.18
Beilstein:
272788
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

mp

>300 °C (lit.)

semiconductor properties

N-type (mobility=0.003 cm2/V·s)

SMILES string

O=C1OC(=O)c2ccc3C(=O)OC(=O)c4ccc1c2c34

InChI

1S/C14H4O6/c15-11-5-1-2-6-10-8(14(18)20-12(6)16)4-3-7(9(5)10)13(17)19-11/h1-4H

InChI key

YTVNOVQHSGMMOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4,5,8-Naphthalenetetracarboxylic dianhydride (NTCDA) is a tetrafunctional monomer that can be used as an electron transporting material in organic electronics.

Application

An n-channel organic semiconductor.
NTCDA can be used in the fabrication of a variety of devices such as fuel cells, thin film transistors (OTFTs), lithium ion batteries, and organic photovoltaics (OPV).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical performance of sol-gel synthesized LiFePO4 in lithium batteries
Hu Y, et al.
Journal of the Electrochemical Society, 151(8), A1279-A1285 (2004)
Mohammad A Jafar Mazumder et al.
Scientific reports, 10(1), 7049-7049 (2020-04-29)
Sulfonated homo and co- polyimide (sPI) were synthesized with new compositional ratios, and used as additives (0.5 wt%, 0.75 wt%, and 1.0 wt%) to prepare blend membranes with polysulfone (PSf). Flat sheet membranes for ultrafiltration (UF) were casted using the phase inversion technique.
Efficient p-i-n type organic solar cells incorporating 1, 4, 5, 8-naphthalenetetracarboxylic dianhydride as transparent electron transport material
Falkenberg C, et al.
Journal of Applied Physics, 104(3), 034506-034506 (2008)
Multi-parameter gas sensors based on organic thin-film-transistors
Torsi L, et al.
Sensors and Actuators B, Chemical, 67(3), 312-316 (2000)
Sulfonated polyimides with flexible aliphatic side chains for polymer electrolyte fuel cells
Yin Y, et al.
Journal of Membrane Science , 367(1-2), 211-219 (2011)

Articles

Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service