Skip to Content
Merck
All Photos(1)

Key Documents

295388

Sigma-Aldrich

Hexafluoropropene

≥99%

Synonym(s):

Hexafluoropropylene, Perfluoropropene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CF=CF2
CAS Number:
Molecular Weight:
150.02
Beilstein:
1705671
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

bp

−28 °C (lit.)

mp

−153 °C (lit.)

SMILES string

F\C(F)=C(/F)C(F)(F)F

InChI

1S/C3F6/c4-1(2(5)6)3(7,8)9

InChI key

HCDGVLDPFQMKDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Supplied in a Sure/Pac cylinder and has a brass needle valve with a male 1/4" NPTF outlet thread installed. Before using the cylinder, ensure that the valve is closed, then remove the galvanized steel hex cap that seals the outlet valve

Compatible with the following:

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Press. Gas Liquefied gas - STOT RE 2 - STOT SE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jenny W Reboredo et al.
Advanced healthcare materials, 5(17), 2191-2198 (2016-05-18)
Cartilage degeneration is the major cause of chronic pain, lost mobility, and reduced quality of life for over estimated 150 million osteoarthritis sufferers worldwide. Despite intensive research, none of the available therapies can restore the hyaline cartilage surface beyond just
Xiuzhi Tian et al.
Journal of hazardous materials, 153(1-2), 128-135 (2007-09-22)
In this study, poly(vinylidene fluoride-co-hexafluoropropene) (PVDF-HFP) with low crystallinity was applied as the membrane material for pervaporative separating ethyl acetate (EtAc) from its aqueous solutions. The drying conditions during membrane fabrication by means of casting the PVDF-HFP solution dominated the
Rafał Loska et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(8), 2577-2589 (2008-01-15)
Hexafluoropropene reacts with aromatic azine N-oxides under mild conditions to produce fluorides of 2-heteroarylperfluoropropionic acids. The reaction proceeds as 1,3-dipolar cycloaddition followed by spontaneous scission of the N--O bond in the isoxazolidine ring and elimination of HF. When the reaction
M Koob et al.
Drug metabolism and disposition: the biological fate of chemicals, 18(6), 911-916 (1990-11-01)
We investigated the metabolism of hexafluoropropene, a nephrotoxic fluoroalkene, in rat liver and kidney subcellular fractions and in rats in vivo. Incubation of hexafluoropropene (1 mM) with microsomes and cytosol in the presence of glutathione (GSH) yielded S-(1,2,3,3,3-pentafluoropropenyl)glutathione (PPFG) and
Effects of inhaled chlorotrifluoroethylene and hexafluoropropene on the rat kidney.
C L Potter et al.
Toxicology and applied pharmacology, 59(3), 431-440 (1981-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service