Skip to Content
Merck
All Photos(1)

Key Documents

294675

Sigma-Aldrich

trans-2,cis-6-Nonadienal

95%

Synonym(s):

Violet leaf aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=CHCH2CH2CH=CHCHO
CAS Number:
Molecular Weight:
138.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.474 (lit.)

bp

94-95 °C/18 mmHg (lit.)

density

0.86 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

[H]C(=O)C(\[H])=C(/[H])CC\C([H])=C(\[H])CC

InChI

1S/C9H14O/c1-2-3-4-5-6-7-8-9-10/h3-4,7-9H,2,5-6H2,1H3/b4-3-,8-7+

InChI key

HZYHMHHBBBSGHB-ODYTWBPASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Unialgal cultures of various Synurophycean species for the occurrence of trans-2,cis-6-nonadienal have been investigated.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

181.4 °F

Flash Point(C)

83 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Production of the taste/odor-causing compound, trans-2, cis-6-nonadienal, within the synurophyceae.
Wee JL, et al.
Journal of Applied Phycology, 6(4), 365-369 (1994)
Michael Czerny et al.
Journal of agricultural and food chemistry, 50(23), 6835-6840 (2002-10-31)
An investigation on the odor-active compounds of wholemeal (WWF) and white wheat flour (WF 550) by aroma extract dilution analysis (AEDA) and by quantitative studies using stable isotope dilution assays (SIDA) revealed a significant number of odor-active compounds, such as
U Dittberner et al.
Mutation research, 335(3), 259-265 (1995-12-01)
The genotoxic effects of the 2-alkenals crotonaldehyde, 2-trans-hexenal and 2-trans-6-cis-nonadienal were studied by cytogenetic methods, analyzing frequencies of sister-chromatid-exchanges, numerical and structural chromosome aberrations and micronucleus induction in human blood lymphocytes and cells of the permanent Namalva line. Crotonaldehyde and
Cheryl Palma-Harris et al.
Journal of agricultural and food chemistry, 50(17), 4875-4877 (2002-08-09)
The ability of nonacidified, refrigerated pickled cucumbers to produce the fresh cucumber flavor impact compounds (E,Z)-2,6-nonadienal and (E)-2-nonenal declined during storage. Production of these compounds decreased as the pH of refrigerated cucumbers was reduced. Despite the fact that the concentrations
Pinar Omür-Ozbek et al.
Environmental science & technology, 45(2), 468-473 (2010-12-15)
Modeling of human exposure to aqueous algal odorants geosmin (earthy), 2-methylisoborneol (musty), and (trans,cis)-2,6-nonadienal (cucumber, fishy), and the solvent trichloroethylene (sweet chemical), was investigated to improve the understanding of water-air transfer by including humans as sensors to detect contaminants. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service