Skip to Content
Merck
All Photos(3)

Key Documents

289418

Sigma-Aldrich

Indium(III) oxide

99.99% trace metals basis

Synonym(s):

Diindium trioxide, Indium sesquioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
In2O3
CAS Number:
Molecular Weight:
277.63
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 25 °C)

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: indium

density

7.18 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[In]O[In]=O

InChI

1S/2In.3O

InChI key

SHTGRZNPWBITMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indium(III)oxide, also known as indium sesquioxide or In2O3. It is athermally stable compound, suitable for use in glass, optic, and ceramic applications.Indium oxide is also commonly doped with tin oxide (SnO2) to produce indium tinoxide (ITO), which is used in transparent thin conductive films for displays,energy-efficient windows, and photovoltaics. Indium oxide-based catalysts are alsoused in various catalytic applications including CO2 hydrogenation, directconversion of syngas into light olefins, and conversion of meso-epoxide intochiral β-amino alcohols.

Application

  • Comparative analysis on application conditions of indium (III) oxide-reinforced glasses in nuclear waste management and source transportation: A Monte Carlo study: This research explores the use of indium (III) oxide-reinforced glass for radioactive waste containment, highlighting its effectiveness and potential in nuclear waste management (ALMisned et al., 2023).
  • Double-shelled hollow rods assembled from nitrogen/sulfur-codoped carbon coated indium oxide nanoparticles as excellent photocatalysts: Discusses the synthesis and application of indium oxide nanoparticles in photocatalysis, demonstrating significant enhancements in environmental cleanup technologies (Sun et al., 2019).
  • Black indium oxide a photothermal CO2 hydrogenation catalyst: Investigates black indium oxide for its use in photocatalytic CO2 reduction, a critical process for sustainable energy and chemical synthesis (Wang et al., 2020).
  • Material proposal for 2D indium oxide: This study proposes two-dimensional indium oxide, discussing its material characteristics and potential applications in electronics and optoelectronics (Kakanakova-Georgieva et al., 2021).
  • Purification of indium by solvent extraction with undiluted ionic liquids: Examines the processes of extracting and purifying indium using green chemistry approaches, contributing to more sustainable practices in materials processing (Deferm et al., 2016).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yiping Fang et al.
Langmuir : the ACS journal of surfaces and colloids, 27(23), 14091-14095 (2011-10-21)
In(2)O(3)@SiO(2) core-shell nanoparticles were prepared using an organic solution synthesis approach and reverse-microemulsion technique. In order to explore the availability of various silica encapsulations, a partial phase diagram for this ternary system consisting of hexane/cyclohexane (1:29 wt), surfactant (polyoxyethylene(5)nonylphenyl ether
Ilan Jen-La Plante et al.
Small (Weinheim an der Bergstrasse, Germany), 9(1), 56-60 (2012-11-06)
Nano popcorn: a new formation mechanism for the synthesis of hollow metal oxide nanoparticles through a melt fracture mechanism. The hollow nanoparticles are formed via brittle fracture following the generation of tensile stresses arising due to liquid-phase thermal expansion of
Wen-Ku Chang et al.
Nanomedicine : nanotechnology, biology, and medicine, 8(5), 609-617 (2011-10-29)
Antibacterial activity of photocatalytic substrates is primarily induced by ultraviolet light irradiation. Visible light-responsive photocatalysts were recently discovered, offering greater opportunity to use photocatalysts as disinfectants in our living environment. The development of antibacterial photocatalysts, however, has mainly focused on
Kelvin H L Zhang et al.
ACS nano, 6(8), 6717-6729 (2012-06-30)
The growth of In(2)O(3) on cubic Y-stabilized ZrO(2)(001) by molecular beam epitaxy leads to formation of nanoscale islands which may tilt relative to the substrate in order to help accommodate the 1.7% tensile mismatch between the epilayer and the substrate.
Jiefu Yin et al.
Inorganic chemistry, 51(12), 6529-6536 (2012-06-06)
We report here for the first time the hollow, metastable, single-crystal, rhombohedral In(2)O(3) (rh-In(2)O(3)) nanocrystals synthesized by annealing solvothermally prepared InOOH solid nanocrystals under ambient pressure at 400 °C, through a mechanism of the Kirkendall effect, in which pore formation

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service