Skip to Content
Merck
All Photos(1)

Key Documents

307874

Sigma-Aldrich

Scandium(III) oxide

99.9% trace rare earth metals basis

Synonym(s):

Discandium trioxide, Scandia, Scandium oxide, Scandium trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sc2O3
CAS Number:
Molecular Weight:
137.91
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99.9% trace rare earth metals basis

form

powder

reaction suitability

core: scandium
reagent type: catalyst

SMILES string

O=[Sc]O[Sc]=O

InChI

1S/3O.2Sc

InChI key

HYXGAEYDKFCVMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Scandium(III) oxide is a high-melting rare earth oxide that can be used as a catalyst in the dehydration of terminal diols. Furthermore, it is used as a starting material for the preparation of other scandium compounds.

Application

Scandium(III) oxide can be used as a catalyst for the dehydration of amino alcohols to unsaturated amines. It can also be used as a precursor to prepare scandium(III) trifluoromethanesulfonate, which can be used to catalyze the nitration of aromatic compounds.
Suitable for vacuum deposition applications

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dehydration of 5-amino-1-pentanol over rare earth oxides
Ohta K, et al.
Applied Catalysis A: General, 517, 73-80 (2016)
Scandium (III) trifluoromethanesulfonate catalyzed aromatic nitration with inorganic nitrates and acetic anhydride
Kawada A, et al.
Chemical & Pharmaceutical Bulletin, 50 (2002)
Vapor-phase catalytic dehydration of terminal diols
Abe K, et al.
Catalysis Today, 164, 419-424 (2011)
R Soulard et al.
Optics express, 18(11), 11173-11180 (2010-07-01)
We report on the measurements of near-UV excited-state absorption (ESA) spectra and refractive index changes (RICs) in the two ytterbium doped laser crystals Yb:Lu2O3 and Yb:Sc2O3. ESA is assigned to ligand-to-metal charge transfer (LMCT) absorption transitions and RICs to the
[Industrial hygiene in scandium oxide manufacture].
B G Feĭgin
Gigiena truda i professional'nye zabolevaniia, (4)(4), 39-40 (1985-04-01)

Articles

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service