Skip to Content
Merck
All Photos(1)

Documents

857335

Sigma-Aldrich

(1S,2R)-(+)-Ephedrine hydrochloride

99%

Synonym(s):

(1S,2R)-(+)-α-(1-Methylaminoethyl)benzyl alcohol hydrochloride, (1S,2R)-(+)-2-Methylamino-1-phenyl-1-propanol hydrochloride, D-α-(1-Methylaminoethyl)benzyl alcohol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH[CH(NHCH3)CH3]OH · HCl
CAS Number:
Molecular Weight:
201.69
Beilstein:
6118541
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

crystals

optical activity

[α]23/D +34.3°, c = 5 in H2O

mp

218-220 °C (lit.)

SMILES string

Cl[H].CN[C@H](C)[C@@H](O)c1ccccc1

InChI

1S/C10H15NO.ClH/c1-8(11-2)10(12)9-6-4-3-5-7-9;/h3-8,10-12H,1-2H3;1H/t8-,10-;/m1./s1

InChI key

BALXUFOVQVENIU-GHXDPTCOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ephedrines are quite significant components in asymmetric synthesis; they are commonly used as key components in the preparation of heterocyclic compounds. They are also used as reagents/ catalytic substances or ligands for the synthesis of aryl/heteroaryl derivatives and organometallic complexes.

Application

(1S,2R)-(+)-Ephedrine hydrochloride may be used in the preparation of (R)-methcathinone via permanganate method of oxidation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

An efficient synthesis of aziridines from ephedrines
Cruz A, et al.
Tetrahedron Asymmetry, 21(8), 909-913 (2010)
Metal-Drug Interactions: Synthesis and Spectroscopic Characteristics, Surface Morphology, and Pharmacological Activity of Ephedrine-HCl Complexes with Mo (V), Nb (V), Ga (III), and Ge (IV)
El-Habeeb AA and Refat MS
Russ. J. Gen. Chem., 88(10), 2163-2169 (2018)
Efficient synthesis of cis-thiazolidinethiones derived from ephedrines
Cruz A, et al.
Tetrahedron Asymmetry, 22(4), 394-398 (2011)
Chiral identification and determination of ephedrine, pseudoephedrine, methamphetamine and metecathinone by gas chromatography and nuclear magnetic resonance.
LeBelle MJ, et al.
Forensic Science International, 71(3), 215-223 (1995)
Noel Kong et al.
Journal of separation science, 37(21), 3015-3023 (2014-08-22)
A sensitive hydrophilic interaction liquid chromatography coupled with tandem mass spectrometry method was developed and validated for the simultaneous detection and quantification of etilefrine and oxilofrine in equine blood plasma and urine. The method is highly sensitive and specific with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service