Skip to Content
Merck
All Photos(1)

Key Documents

28682

Sigma-Aldrich

cis-Cyclobutane-1,2-dicarboxylic acid

≥97.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O4
CAS Number:
Molecular Weight:
144.13
Beilstein:
2047669
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (T)

form

solid

mp

133-139 °C

functional group

carboxylic acid

SMILES string

OC(=O)[C@@H]1CC[C@@H]1C(O)=O

InChI

1S/C6H8O4/c7-5(8)3-1-2-4(3)6(9)10/h3-4H,1-2H2,(H,7,8)(H,9,10)/t3-,4+

InChI key

SUSAGCZZQKACKE-ZXZARUISSA-N

General description

The interaction of the carboxyl groups in the fragmentation of the molecular ions of cis-cyclobutane-1,2-dicarboxylic acid helps to distinguish it from trans-cyclobutane-1,2-dicarboxylic acid.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benoit F and Holmes JL.
Org. Mass Spectrom., 6(5), 541-548 (1972)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service