Skip to Content
Merck
All Photos(1)

Key Documents

206075

Sigma-Aldrich

Potassium tetrachloroplatinate(II)

98%

Synonym(s):

Potassium platinum(II) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2PtCl4
CAS Number:
Molecular Weight:
415.09
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid

grade

for analytical purposes

Assay

98%

form

solid

density

3.38 g/mL at 25 °C (lit.)

SMILES string

[K+].[K+].Cl[Pt--](Cl)(Cl)Cl

InChI

1S/4ClH.2K.Pt/h4*1H;;;/q;;;;2*+1;+2/p-4

InChI key

RVRDLMCWUILSAH-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Potassium tetrachloroplatinate(II) is a ruby-red crystalline solid and can be prepared by reacting potassium hexachloroplatinate(IV) with cuprous chloride. It is mainly used as a reagent and inorganic catalyst in various chemical reactions. Its application includes the synthesis of other platinum compounds, organic transformations, and electroplating of metals. It is also a potential antitumor agent.

Application

Potassium tetrachloroplatinate(II) can be used as:
  • A precursor to synthesize CO-tolerant catalyst for hydrogen-air fuel cells.
  • An active and stable catalyst for selective methane oxidation.
  • As a starting material to synthesize high-performance Pt-based phosphorescent materials for electroluminescent devices.

Features and Benefits

Reacts with bis(dithiolates) to form metal-bis(dithiolates) with applications in laser Q-switch materials, liquid crystals, optical CD recording media, bar code materials and superconductivity. Reacts with capping polymers to form shape-control colloidal platinum nanoparticles for catalysis and photocatalysts.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of Pt-HxMoO3 catalysts for CO-tolerant PEMFCs
Kirill A. Kurdin, et al.
Catalysis Today, 388-389, 147-157 (2022)
Raj G
Advances in Inorganic Chemistry null
In search of high-performance platinum (II) phosphorescent materials for the fabrication of red electroluminescent devices
Jakka Kavitha, et al.
Advanced Functional Materials , 15, 223-229 (2005)
Kyubin Shim et al.
Scientific reports, 9(1), 894-894 (2019-01-31)
Bimetallic nanoparticles (NPs) have aroused interest in various fields because of their synergetic and unique properties. Among those nanoparticles, we strategically approached and synthesized Au@Pt NPs via the sonochemical method with different molar ratios (e.g. 3:7, 5:5, and 7:3) of
The Biological Activity of Zeise's Salt and its Derivatives
Meieranz S, et al.
Angewandte Chemie (International Edition in English), 54(9), 2834-2837 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service